Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67294884-67295157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD19
Entrez Gene ID
146212
Gene Name
potassium channel tetramerization domain containing 19
Gene Aliases
-
UniGene
Hs.299127
Gene Chromosome Location
Chr.16: 67289490-67326758 on Build GRCh38
Transcripts
NM_001100915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7267870 c.1392-8G>A p.? Chr.16: 67295064-67295064 on Build GRCh38
1679181 c.1393G>A p.E465K Chr.16: 67295055-67295055 on Build GRCh38
5718747 c.1423G>A p.E475K Chr.16: 67295025-67295025 on Build GRCh38
6464444 c.1431C>T p.H477H Chr.16: 67295017-67295017 on Build GRCh38
6079523 c.1465G>A p.E489K Chr.16: 67294983-67294983 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749842965 Chr.16: 67294953-67294953 on Build GRCh38
rs780787043 Chr.16: 67294952-67294952 on Build GRCh38
rs775860778 Chr.16: 67294947-67294948 on Build GRCh38
rs918350053 Chr.16: 67295120-67295120 on Build GRCh38
rs755589250 Chr.16: 67295092-67295092 on Build GRCh38
rs751805831 Chr.16: 67295134-67295134 on Build GRCh38
rs952786515 Chr.16: 67295026-67295026 on Build GRCh38
rs751492532 Chr.16: 67294934-67294934 on Build GRCh38
rs770236578 Chr.16: 67294968-67294971 on Build GRCh38
rs541694257 Chr.16: 67294928-67294928 on Build GRCh38
rs776962794 Chr.16: 67295060-67295060 on Build GRCh38
rs369286414 Chr.16: 67295106-67295106 on Build GRCh38
rs531423837 Chr.16: 67295088-67295088 on Build GRCh38
rs187644248 Chr.16: 67295070-67295070 on Build GRCh38
rs929704146 Chr.16: 67295133-67295133 on Build GRCh38
rs761821556 Chr.16: 67295084-67295084 on Build GRCh38
rs908624390 Chr.16: 67295048-67295048 on Build GRCh38
rs976551445 Chr.16: 67295110-67295110 on Build GRCh38
rs750015847 Chr.16: 67295090-67295090 on Build GRCh38
rs772395505 Chr.16: 67295011-67295011 on Build GRCh38
rs779196947 Chr.16: 67295008-67295008 on Build GRCh38
rs773358045 Chr.16: 67295012-67295012 on Build GRCh38
rs762891138 Chr.16: 67295068-67295068 on Build GRCh38
rs373158408 Chr.16: 67295113-67295113 on Build GRCh38
rs553717381 Chr.16: 67294932-67294932 on Build GRCh38
rs757095239 Chr.16: 67294935-67294935 on Build GRCh38
rs557828022 Chr.16: 67294975-67294975 on Build GRCh38
rs771350240 Chr.16: 67295037-67295037 on Build GRCh38
rs747050776 Chr.16: 67295030-67295030 on Build GRCh38
rs201322927 Chr.16: 67295082-67295082 on Build GRCh38
rs767342488 Chr.16: 67295086-67295086 on Build GRCh38
rs753506480 Chr.16: 67295062-67295064 on Build GRCh38
rs749689690 Chr.16: 67294986-67294986 on Build GRCh38
rs200312357 Chr.16: 67294988-67294988 on Build GRCh38
rs779675369 Chr.16: 67294977-67294977 on Build GRCh38
rs374183561 Chr.16: 67294954-67294954 on Build GRCh38
rs760419648 Chr.16: 67294960-67294961 on Build GRCh38

View Full Product Details