Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236226406-236226667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERO1B
Entrez Gene ID
56605
Gene Name
endoplasmic reticulum oxidoreductase 1 beta
Gene Aliases
ERO1LB, Ero1beta
UniGene
Hs.558519
Gene Chromosome Location
Chr.1: 236215122-236282039 on Build GRCh38
Transcripts
NM_019891.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
372591 c.800T>A p.L267* Chr.1: 236226652-236226652 on Build GRCh38
7279052 c.856C>T p.R286C Chr.1: 236226465-236226465 on Build GRCh38
1196854 c.903G>T p.K301N Chr.1: 236226418-236226418 on Build GRCh38
5978263 c.809C>A p.T270N Chr.1: 236226512-236226512 on Build GRCh38
4029802 c.831A>G p.G277G Chr.1: 236226490-236226490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563572029 Chr.1: 236226512-236226512 on Build GRCh38
rs369252480 Chr.1: 236226499-236226499 on Build GRCh38
rs904042171 Chr.1: 236226583-236226583 on Build GRCh38
rs373246490 Chr.1: 236226483-236226483 on Build GRCh38
rs780062804 Chr.1: 236226465-236226465 on Build GRCh38
rs763983564 Chr.1: 236226610-236226611 on Build GRCh38
rs907105108 Chr.1: 236226453-236226453 on Build GRCh38
rs35475164 Chr.1: 236226560-236226560 on Build GRCh38
rs951855227 Chr.1: 236226542-236226542 on Build GRCh38
rs757075087 Chr.1: 236226440-236226440 on Build GRCh38
rs566376855 Chr.1: 236226613-236226613 on Build GRCh38
rs774133514 Chr.1: 236226537-236226537 on Build GRCh38
rs759329682 Chr.1: 236226544-236226544 on Build GRCh38
rs377302108 Chr.1: 236226597-236226597 on Build GRCh38
rs768998792 Chr.1: 236226477-236226477 on Build GRCh38
rs760253373 Chr.1: 236226603-236226603 on Build GRCh38
rs1014610495 Chr.1: 236226580-236226580 on Build GRCh38
rs984525653 Chr.1: 236226567-236226567 on Build GRCh38
rs536066281 Chr.1: 236226442-236226442 on Build GRCh38
rs776595450 Chr.1: 236226633-236226633 on Build GRCh38
rs772121664 Chr.1: 236226464-236226464 on Build GRCh38
rs775341431 Chr.1: 236226559-236226560 on Build GRCh38
rs760454691 Chr.1: 236226608-236226609 on Build GRCh38
rs10924789 Chr.1: 236226626-236226626 on Build GRCh38
rs753525753 Chr.1: 236226433-236226433 on Build GRCh38
rs757019994 Chr.1: 236226628-236226628 on Build GRCh38
rs746134572 Chr.1: 236226461-236226461 on Build GRCh38
rs767205158 Chr.1: 236226561-236226561 on Build GRCh38
rs748496400 Chr.1: 236226498-236226498 on Build GRCh38
rs747113751 Chr.1: 236226466-236226466 on Build GRCh38

View Full Product Details