Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165482155-165482408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPE
Entrez Gene ID
1363
Gene Name
carboxypeptidase E
Gene Aliases
CPH
UniGene
Hs.75360
Gene Chromosome Location
Chr.4: 165378942-165498547 on Build GRCh38
Transcripts
NM_001873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5923665 c.682G>A p.E228K Chr.4: 165482251-165482251 on Build GRCh38
5709153 c.700C>T p.H234Y Chr.4: 165482269-165482269 on Build GRCh38
4856017 c.731C>A p.S244Y Chr.4: 165482300-165482300 on Build GRCh38
4123314 c.751G>T p.D251Y Chr.4: 165482320-165482320 on Build GRCh38
1695210 c.712G>A p.D238N Chr.4: 165482281-165482281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750717332 Chr.4: 165482336-165482336 on Build GRCh38
rs868811858 Chr.4: 165482314-165482314 on Build GRCh38
rs777794895 Chr.4: 165482229-165482229 on Build GRCh38
rs376055726 Chr.4: 165482353-165482353 on Build GRCh38
rs77809551 Chr.4: 165482293-165482293 on Build GRCh38
rs972676357 Chr.4: 165482204-165482204 on Build GRCh38
rs774863176 Chr.4: 165482279-165482279 on Build GRCh38
rs765565656 Chr.4: 165482322-165482322 on Build GRCh38
rs756009956 Chr.4: 165482215-165482215 on Build GRCh38
rs771676991 Chr.4: 165482271-165482271 on Build GRCh38
rs371049668 Chr.4: 165482288-165482288 on Build GRCh38
rs762207857 Chr.4: 165482313-165482313 on Build GRCh38
rs80117208 Chr.4: 165482352-165482352 on Build GRCh38
rs1035007938 Chr.4: 165482321-165482321 on Build GRCh38
rs745606793 Chr.4: 165482373-165482373 on Build GRCh38
rs758640131 Chr.4: 165482228-165482228 on Build GRCh38
rs34516004 Chr.4: 165482272-165482272 on Build GRCh38
rs140559965 Chr.4: 165482351-165482351 on Build GRCh38
rs145394460 Chr.4: 165482354-165482354 on Build GRCh38
rs779987140 Chr.4: 165482193-165482193 on Build GRCh38
rs200369500 Chr.4: 165482306-165482306 on Build GRCh38
rs202187157 Chr.4: 165482369-165482369 on Build GRCh38
rs193118404 Chr.4: 165482265-165482265 on Build GRCh38
rs971092327 Chr.4: 165482260-165482260 on Build GRCh38
rs763083392 Chr.4: 165482338-165482338 on Build GRCh38
rs200405029 Chr.4: 165482202-165482202 on Build GRCh38
rs377643898 Chr.4: 165482232-165482232 on Build GRCh38
rs764611490 Chr.4: 165482303-165482303 on Build GRCh38
rs745661164 Chr.4: 165482366-165482366 on Build GRCh38
rs892935017 Chr.4: 165482214-165482214 on Build GRCh38
rs914770769 Chr.4: 165482342-165482342 on Build GRCh38
rs770817173 Chr.4: 165482245-165482245 on Build GRCh38
rs199819898 Chr.4: 165482340-165482340 on Build GRCh38

View Full Product Details