Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194342485-194342730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPN2
Entrez Gene ID
1370
Gene Name
carboxypeptidase N subunit 2
Gene Aliases
ACBP
UniGene
Hs.528368
Gene Chromosome Location
Chr.3: 194339765-194351387 on Build GRCh38
Transcripts
NM_001080513.3 NM_001291988.1 XM_005269280.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1694005 c.155A>C p.K52T Chr.3: 194342548-194342548 on Build GRCh38
6634618 c.28A>G p.T10A Chr.3: 194342675-194342675 on Build GRCh38
3781793 c.98T>G p.V33G Chr.3: 194342605-194342605 on Build GRCh38
3333914 c.6C>T p.L2L Chr.3: 194342697-194342697 on Build GRCh38
5792308 c.154A>G p.K52E Chr.3: 194342549-194342549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs9845073 Chr.3: 194342615-194342615 on Build GRCh38
rs867589718 Chr.3: 194342613-194342613 on Build GRCh38
rs763128530 Chr.3: 194342568-194342568 on Build GRCh38
rs768470859 Chr.3: 194342633-194342633 on Build GRCh38
rs778326304 Chr.3: 194342550-194342550 on Build GRCh38
rs143058335 Chr.3: 194342524-194342524 on Build GRCh38
rs146166274 Chr.3: 194342551-194342551 on Build GRCh38
rs148539182 Chr.3: 194342689-194342689 on Build GRCh38
rs749508351 Chr.3: 194342653-194342653 on Build GRCh38
rs753497669 Chr.3: 194342590-194342590 on Build GRCh38
rs74820225 Chr.3: 194342559-194342559 on Build GRCh38
rs768067685 Chr.3: 194342576-194342576 on Build GRCh38
rs921894077 Chr.3: 194342579-194342579 on Build GRCh38
rs779920381 Chr.3: 194342616-194342616 on Build GRCh38
rs370375539 Chr.3: 194342652-194342652 on Build GRCh38
rs140187491 Chr.3: 194342661-194342661 on Build GRCh38
rs142022933 Chr.3: 194342674-194342674 on Build GRCh38
rs377176546 Chr.3: 194342560-194342560 on Build GRCh38
rs777423874 Chr.3: 194342540-194342540 on Build GRCh38
rs761048954 Chr.3: 194342578-194342578 on Build GRCh38
rs982803111 Chr.3: 194342572-194342572 on Build GRCh38
rs772298855 Chr.3: 194342678-194342678 on Build GRCh38
rs780903007 Chr.3: 194342651-194342651 on Build GRCh38
rs138306434 Chr.3: 194342609-194342609 on Build GRCh38
rs62288099 Chr.3: 194342571-194342571 on Build GRCh38
rs150511099 Chr.3: 194342577-194342577 on Build GRCh38
rs753175943 Chr.3: 194342523-194342523 on Build GRCh38
rs146122822 Chr.3: 194342622-194342622 on Build GRCh38
rs568207489 Chr.3: 194342655-194342655 on Build GRCh38
rs774626597 Chr.3: 194342570-194342570 on Build GRCh38
rs748914809 Chr.3: 194342547-194342547 on Build GRCh38
rs373784213 Chr.3: 194342693-194342693 on Build GRCh38
rs771025061 Chr.3: 194342569-194342569 on Build GRCh38
rs372991840 Chr.3: 194342556-194342556 on Build GRCh38
rs772064450 Chr.3: 194342555-194342555 on Build GRCh38
rs761125023 Chr.3: 194342700-194342700 on Build GRCh38
rs567975075 Chr.3: 194342596-194342596 on Build GRCh38
rs764396263 Chr.3: 194342582-194342582 on Build GRCh38
rs371140524 Chr.3: 194342548-194342548 on Build GRCh38
rs756909009 Chr.3: 194342595-194342595 on Build GRCh38
rs1024748699 Chr.3: 194342520-194342520 on Build GRCh38

View Full Product Details