Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 45134258-45134485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP3
Entrez Gene ID
2287
Gene Name
FK506 binding protein 3
Gene Aliases
FKBP-25, FKBP-3, FKBP25, PPIase
UniGene
Hs.509226
Gene Chromosome Location
Chr.14: 45115599-45134529 on Build GRCh38
Transcripts
NM_002013.3 XM_011536565.2 XM_017021092.1 XM_017021093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6664258 c.63C>T p.P21P Chr.14: 45134394-45134394 on Build GRCh38
6075800 c.18A>T p.P6P Chr.14: 45134439-45134439 on Build GRCh38
1369842 c.79A>G p.K27E Chr.14: 45134378-45134378 on Build GRCh38
6583291 c.61C>T p.P21S Chr.14: 45134396-45134396 on Build GRCh38
6001384 c.63C>A p.P21P Chr.14: 45134394-45134394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753414901 Chr.14: 45134330-45134330 on Build GRCh38
rs748060039 Chr.14: 45134440-45134440 on Build GRCh38
rs754854617 Chr.14: 45134350-45134350 on Build GRCh38
rs752763261 Chr.14: 45134319-45134319 on Build GRCh38
rs982123640 Chr.14: 45134326-45134326 on Build GRCh38
rs757697121 Chr.14: 45134330-45134330 on Build GRCh38
rs754193578 Chr.14: 45134328-45134328 on Build GRCh38
rs775623307 Chr.14: 45134410-45134410 on Build GRCh38
rs28366026 Chr.14: 45134421-45134421 on Build GRCh38
rs777622667 Chr.14: 45134444-45134444 on Build GRCh38
rs1046582893 Chr.14: 45134414-45134414 on Build GRCh38
rs758063895 Chr.14: 45134434-45134434 on Build GRCh38
rs188886335 Chr.14: 45134397-45134397 on Build GRCh38
rs747898326 Chr.14: 45134377-45134377 on Build GRCh38
rs576960039 Chr.14: 45134293-45134293 on Build GRCh38
rs757296457 Chr.14: 45134430-45134430 on Build GRCh38
rs765910800 Chr.14: 45134465-45134465 on Build GRCh38
rs776899510 Chr.14: 45134449-45134449 on Build GRCh38
rs764753048 Chr.14: 45134325-45134325 on Build GRCh38
rs781112541 Chr.14: 45134368-45134368 on Build GRCh38
rs374796242 Chr.14: 45134320-45134320 on Build GRCh38
rs756087507 Chr.14: 45134443-45134443 on Build GRCh38
rs746074324 Chr.14: 45134391-45134391 on Build GRCh38
rs772252908 Chr.14: 45134394-45134394 on Build GRCh38
rs377237372 Chr.14: 45134298-45134298 on Build GRCh38
rs757731413 Chr.14: 45134395-45134396 on Build GRCh38
rs765674278 Chr.14: 45134431-45134431 on Build GRCh38
rs750791403 Chr.14: 45134432-45134432 on Build GRCh38
rs149780967 Chr.14: 45134437-45134437 on Build GRCh38
rs139680673 Chr.14: 45134379-45134379 on Build GRCh38
rs115291634 Chr.14: 45134445-45134445 on Build GRCh38
rs111523571 Chr.14: 45134384-45134384 on Build GRCh38
rs767615899 Chr.14: 45134302-45134302 on Build GRCh38
rs200895253 Chr.14: 45134345-45134345 on Build GRCh38
rs569957294 Chr.14: 45134401-45134401 on Build GRCh38
rs909783158 Chr.14: 45134424-45134424 on Build GRCh38
rs367659655 Chr.14: 45134338-45134338 on Build GRCh38
rs762376485 Chr.14: 45134464-45134464 on Build GRCh38
rs536915344 Chr.14: 45134439-45134439 on Build GRCh38
rs1013264249 Chr.14: 45134359-45134359 on Build GRCh38
rs746560206 Chr.14: 45134349-45134349 on Build GRCh38
rs970836634 Chr.14: 45134304-45134304 on Build GRCh38
rs964893414 Chr.14: 45134453-45134453 on Build GRCh38
rs1024166051 Chr.14: 45134294-45134294 on Build GRCh38
rs929311192 Chr.14: 45134297-45134297 on Build GRCh38
rs758034623 Chr.14: 45134342-45134342 on Build GRCh38
rs923274274 Chr.14: 45134292-45134293 on Build GRCh38
rs775575091 Chr.14: 45134446-45134446 on Build GRCh38
rs760681690 Chr.14: 45134412-45134412 on Build GRCh38
rs976597992 Chr.14: 45134279-45134279 on Build GRCh38
rs765963124 Chr.14: 45134435-45134435 on Build GRCh38
rs534477722 Chr.14: 45134447-45134447 on Build GRCh38
rs576817746 Chr.14: 45134461-45134461 on Build GRCh38
rs762218930 Chr.14: 45134425-45134425 on Build GRCh38

View Full Product Details