Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4738323-4738570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL16
Entrez Gene ID
58191
Gene Name
C-X-C motif chemokine ligand 16
Gene Aliases
CXCLG16, SR-PSOX, SRPSOX
UniGene
Hs.745037
Gene Chromosome Location
Chr.17: 4733529-4739928 on Build GRCh38
Transcripts
NM_001100812.1 NM_022059.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130339 c.306G>A p.G102G Chr.17: 4738460-4738460 on Build GRCh38
6368030 c.313A>T p.K105* Chr.17: 4738453-4738453 on Build GRCh38
4067345 c.349G>A p.D117N Chr.17: 4738417-4738417 on Build GRCh38
5852819 c.346C>T p.L116F Chr.17: 4738420-4738420 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377542126 Chr.17: 4738404-4738404 on Build GRCh38
rs369441846 Chr.17: 4738432-4738432 on Build GRCh38
rs775976461 Chr.17: 4738445-4738445 on Build GRCh38
rs776689689 Chr.17: 4738357-4738357 on Build GRCh38
rs752321474 Chr.17: 4738385-4738385 on Build GRCh38
rs373173069 Chr.17: 4738473-4738473 on Build GRCh38
rs780589545 Chr.17: 4738525-4738525 on Build GRCh38
rs544890776 Chr.17: 4738540-4738540 on Build GRCh38
rs772272880 Chr.17: 4738422-4738422 on Build GRCh38
rs777528672 Chr.17: 4738405-4738405 on Build GRCh38
rs751296517 Chr.17: 4738368-4738368 on Build GRCh38
rs752658376 Chr.17: 4738496-4738496 on Build GRCh38
rs374983118 Chr.17: 4738512-4738512 on Build GRCh38
rs112749067 Chr.17: 4738407-4738407 on Build GRCh38
rs756895767 Chr.17: 4738426-4738426 on Build GRCh38
rs774342387 Chr.17: 4738490-4738490 on Build GRCh38
rs758736278 Chr.17: 4738523-4738523 on Build GRCh38
rs149830899 Chr.17: 4738447-4738447 on Build GRCh38
rs750857845 Chr.17: 4738522-4738522 on Build GRCh38
rs370461293 Chr.17: 4738494-4738494 on Build GRCh38
rs371450795 Chr.17: 4738367-4738367 on Build GRCh38
rs201607328 Chr.17: 4738538-4738538 on Build GRCh38
rs754975512 Chr.17: 4738370-4738370 on Build GRCh38
rs769996795 Chr.17: 4738481-4738481 on Build GRCh38
rs1050997 Chr.17: 4738460-4738460 on Build GRCh38
rs772629335 Chr.17: 4738436-4738436 on Build GRCh38
rs748916923 Chr.17: 4738425-4738425 on Build GRCh38
rs140999028 Chr.17: 4738435-4738435 on Build GRCh38
rs753324129 Chr.17: 4738511-4738511 on Build GRCh38
rs763545891 Chr.17: 4738510-4738510 on Build GRCh38
rs747540761 Chr.17: 4738534-4738534 on Build GRCh38
rs369273133 Chr.17: 4738379-4738379 on Build GRCh38
rs760611199 Chr.17: 4738506-4738506 on Build GRCh38
rs761942667 Chr.17: 4738474-4738474 on Build GRCh38
rs778615024 Chr.17: 4738519-4738519 on Build GRCh38
rs144767148 Chr.17: 4738458-4738458 on Build GRCh38
rs1041840703 Chr.17: 4738420-4738420 on Build GRCh38
rs755652186 Chr.17: 4738389-4738389 on Build GRCh38
rs762042334 Chr.17: 4738362-4738362 on Build GRCh38

View Full Product Details