Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49167113-49167342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC3A
Entrez Gene ID
22862
Gene Name
fibronectin type III domain containing 3A
Gene Aliases
FNDC3, HUGO, bA203I16.1, bA203I16.5
UniGene
Hs.508010
Gene Chromosome Location
Chr.13: 48975241-49209779 on Build GRCh38
Transcripts
NM_001079673.1 NM_001278438.1 NM_014923.4 NR_103528.1 XM_011534997.2 XM_017020440.1 XM_017020441.1 XM_017020442.1 XM_017020443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1706771 c.1034C>G p.A345G Chr.13: 49167300-49167300 on Build GRCh38
6055357 c.978-1G>A p.? Chr.13: 49167243-49167243 on Build GRCh38
4685241 c.1008T>C p.D336D Chr.13: 49167274-49167274 on Build GRCh38
5903843 c.978-6C>T p.? Chr.13: 49167238-49167238 on Build GRCh38
6578567 c.1037A>G p.K346R Chr.13: 49167303-49167303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982629067 Chr.13: 49167309-49167309 on Build GRCh38
rs771093837 Chr.13: 49167288-49167288 on Build GRCh38
rs756882400 Chr.13: 49167248-49167248 on Build GRCh38
rs965562103 Chr.13: 49167184-49167184 on Build GRCh38
rs761021101 Chr.13: 49167205-49167205 on Build GRCh38
rs79948910 Chr.13: 49167197-49167197 on Build GRCh38
rs113025834 Chr.13: 49167302-49167302 on Build GRCh38
rs1018107985 Chr.13: 49167163-49167163 on Build GRCh38
rs929889583 Chr.13: 49167199-49167199 on Build GRCh38
rs780856648 Chr.13: 49167249-49167249 on Build GRCh38
rs976709862 Chr.13: 49167188-49167188 on Build GRCh38
rs983489008 Chr.13: 49167230-49167230 on Build GRCh38
rs762855099 Chr.13: 49167193-49167193 on Build GRCh38
rs1034795155 Chr.13: 49167242-49167242 on Build GRCh38
rs774340147 Chr.13: 49167279-49167279 on Build GRCh38
rs894081391 Chr.13: 49167162-49167162 on Build GRCh38
rs761656616 Chr.13: 49167286-49167286 on Build GRCh38
rs529638075 Chr.13: 49167215-49167215 on Build GRCh38
rs745375746 Chr.13: 49167255-49167255 on Build GRCh38
rs372393975 Chr.13: 49167268-49167268 on Build GRCh38
rs776851008 Chr.13: 49167314-49167314 on Build GRCh38
rs778147778 Chr.13: 49167218-49167218 on Build GRCh38
rs769290582 Chr.13: 49167266-49167266 on Build GRCh38
rs368469510 Chr.13: 49167207-49167207 on Build GRCh38
rs753256933 Chr.13: 49167213-49167213 on Build GRCh38
rs755049465 Chr.13: 49167211-49167211 on Build GRCh38
rs764039806 Chr.13: 49167194-49167194 on Build GRCh38
rs768686708 Chr.13: 49167272-49167272 on Build GRCh38
rs942103582 Chr.13: 49167273-49167273 on Build GRCh38
rs909202961 Chr.13: 49167233-49167233 on Build GRCh38

View Full Product Details