Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42903846-42904119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC
Entrez Gene ID
2538
Gene Name
glucose-6-phosphatase catalytic subunit
Gene Aliases
G6PC1, G6PT, G6Pase, GSD1, GSD1a
UniGene
Hs.212293
Gene Chromosome Location
Chr.17: 42900797-42914433 on Build GRCh38
Transcripts
NM_000151.3 NM_001270397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4449444 c.231-1G>A p.? Chr.17: 42903930-42903930 on Build GRCh38
7184926 c.322A>G p.T108A Chr.17: 42904022-42904022 on Build GRCh38
3189766 c.294C>T p.S98S Chr.17: 42903994-42903994 on Build GRCh38
4381618 c.331A>T p.T111S Chr.17: 42904031-42904031 on Build GRCh38
6891889 c.330G>T p.E110D Chr.17: 42904030-42904030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769235934 Chr.17: 42904020-42904020 on Build GRCh38
rs746797957 Chr.17: 42904090-42904090 on Build GRCh38
rs760002677 Chr.17: 42903902-42903902 on Build GRCh38
rs1801175 Chr.17: 42903947-42903947 on Build GRCh38
rs886052955 Chr.17: 42904026-42904026 on Build GRCh38
rs750886274 Chr.17: 42903939-42903939 on Build GRCh38
rs104894567 Chr.17: 42904028-42904028 on Build GRCh38
rs750796596 Chr.17: 42904046-42904046 on Build GRCh38
rs754394520 Chr.17: 42904055-42904055 on Build GRCh38
rs752348580 Chr.17: 42903916-42903916 on Build GRCh38
rs770516709 Chr.17: 42903882-42903882 on Build GRCh38
rs116797100 Chr.17: 42904082-42904082 on Build GRCh38
rs764936031 Chr.17: 42903913-42903913 on Build GRCh38
rs183285298 Chr.17: 42904060-42904060 on Build GRCh38
rs375219700 Chr.17: 42904004-42904004 on Build GRCh38
rs1022981580 Chr.17: 42903954-42903954 on Build GRCh38
rs780047423 Chr.17: 42903942-42903942 on Build GRCh38
rs748363083 Chr.17: 42903955-42903955 on Build GRCh38
rs755612674 Chr.17: 42903960-42903960 on Build GRCh38
rs142917638 Chr.17: 42903958-42903958 on Build GRCh38
rs777452219 Chr.17: 42903994-42903994 on Build GRCh38
rs1026799180 Chr.17: 42903946-42903946 on Build GRCh38
rs368450665 Chr.17: 42904050-42904050 on Build GRCh38
rs377390234 Chr.17: 42903920-42903920 on Build GRCh38
rs759101908 Chr.17: 42904011-42904011 on Build GRCh38
rs762539496 Chr.17: 42904031-42904031 on Build GRCh38
rs148967047 Chr.17: 42903952-42903952 on Build GRCh38
rs1801176 Chr.17: 42903948-42903948 on Build GRCh38
rs756632286 Chr.17: 42903941-42903941 on Build GRCh38
rs771864998 Chr.17: 42903969-42903969 on Build GRCh38
rs779176162 Chr.17: 42903949-42903949 on Build GRCh38
rs570199529 Chr.17: 42904047-42904047 on Build GRCh38
rs752915213 Chr.17: 42904063-42904063 on Build GRCh38
rs776295479 Chr.17: 42903896-42903896 on Build GRCh38
rs767870143 Chr.17: 42904042-42904042 on Build GRCh38
rs777645597 Chr.17: 42904083-42904083 on Build GRCh38
rs781668568 Chr.17: 42904053-42904053 on Build GRCh38
rs1051317287 Chr.17: 42903870-42903870 on Build GRCh38
rs373908549 Chr.17: 42903919-42903919 on Build GRCh38
rs746885993 Chr.17: 42903995-42903995 on Build GRCh38

View Full Product Details