Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233125705-233125966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5D
Entrez Gene ID
3635
Gene Name
inositol polyphosphate-5-phosphatase D
Gene Aliases
SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
UniGene
Hs.262886
Gene Chromosome Location
Chr.2: 233059967-233207903 on Build GRCh38
Transcripts
NM_001017915.2 NM_005541.4 XM_017004004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4425499 c.459C>A p.T153T Chr.2: 233125854-233125854 on Build GRCh38
6686114 c.499C>T p.R167* Chr.2: 233125894-233125894 on Build GRCh38
720218 c.401C>A p.T134N Chr.2: 233125796-233125796 on Build GRCh38
3579198 c.424G>A p.E142K Chr.2: 233125819-233125819 on Build GRCh38
3579197 c.394C>T p.P132S Chr.2: 233125789-233125789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200834931 Chr.2: 233125865-233125865 on Build GRCh38
rs771155830 Chr.2: 233125879-233125879 on Build GRCh38
rs374974999 Chr.2: 233125821-233125821 on Build GRCh38
rs766394970 Chr.2: 233125836-233125836 on Build GRCh38
rs745349988 Chr.2: 233125860-233125860 on Build GRCh38
rs777393014 Chr.2: 233125768-233125768 on Build GRCh38
rs370172183 Chr.2: 233125850-233125850 on Build GRCh38
rs768572195 Chr.2: 233125869-233125869 on Build GRCh38
rs745814808 Chr.2: 233125791-233125791 on Build GRCh38
rs755661121 Chr.2: 233125939-233125939 on Build GRCh38
rs531680084 Chr.2: 233125854-233125854 on Build GRCh38
rs548356548 Chr.2: 233125855-233125855 on Build GRCh38
rs758115369 Chr.2: 233125767-233125767 on Build GRCh38
rs749957200 Chr.2: 233125935-233125935 on Build GRCh38
rs61752227 Chr.2: 233125884-233125884 on Build GRCh38
rs993668313 Chr.2: 233125906-233125906 on Build GRCh38
rs181242219 Chr.2: 233125882-233125882 on Build GRCh38
rs367778042 Chr.2: 233125750-233125750 on Build GRCh38
rs750595499 Chr.2: 233125726-233125726 on Build GRCh38
rs909335175 Chr.2: 233125842-233125842 on Build GRCh38
rs765244250 Chr.2: 233125887-233125887 on Build GRCh38
rs749177559 Chr.2: 233125868-233125868 on Build GRCh38
rs752579631 Chr.2: 233125898-233125898 on Build GRCh38
rs552232273 Chr.2: 233125837-233125837 on Build GRCh38
rs762834507 Chr.2: 233125797-233125797 on Build GRCh38
rs763411419 Chr.2: 233125895-233125895 on Build GRCh38
rs775464802 Chr.2: 233125795-233125795 on Build GRCh38
rs968593154 Chr.2: 233125807-233125807 on Build GRCh38
rs560060210 Chr.2: 233125818-233125818 on Build GRCh38
rs746606835 Chr.2: 233125779-233125779 on Build GRCh38
rs146631523 Chr.2: 233125790-233125790 on Build GRCh38
rs576374397 Chr.2: 233125730-233125730 on Build GRCh38
rs542008487 Chr.2: 233125737-233125737 on Build GRCh38
rs780891424 Chr.2: 233125933-233125933 on Build GRCh38
rs539975896 Chr.2: 233125809-233125809 on Build GRCh38
rs765772956 Chr.2: 233125843-233125843 on Build GRCh38
rs770352133 Chr.2: 233125786-233125786 on Build GRCh38
rs769686085 Chr.2: 233125864-233125864 on Build GRCh38
rs764464155 Chr.2: 233125902-233125902 on Build GRCh38
rs371488249 Chr.2: 233125755-233125755 on Build GRCh38
rs368153152 Chr.2: 233125846-233125846 on Build GRCh38
rs574989226 Chr.2: 233125926-233125926 on Build GRCh38
rs779197283 Chr.2: 233125758-233125758 on Build GRCh38
rs368806518 Chr.2: 233125930-233125930 on Build GRCh38
rs569172731 Chr.2: 233125851-233125851 on Build GRCh38
rs758777451 Chr.2: 233125847-233125847 on Build GRCh38
rs376452748 Chr.2: 233125922-233125922 on Build GRCh38
rs376568019 Chr.2: 233125878-233125878 on Build GRCh38
rs769747258 Chr.2: 233125793-233125793 on Build GRCh38
rs753207715 Chr.2: 233125894-233125894 on Build GRCh38
rs760881867 Chr.2: 233125829-233125829 on Build GRCh38
rs373679891 Chr.2: 233125724-233125724 on Build GRCh38
rs755468463 Chr.2: 233125756-233125756 on Build GRCh38

View Full Product Details