Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206958459-206958730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCAMR
Entrez Gene ID
83953
Gene Name
Fc fragment of IgA and IgM receptor
Gene Aliases
CD351, FCA/MR, FKSG87
UniGene
Hs.145519
Gene Chromosome Location
Chr.1: 206957955-206970657 on Build GRCh38
Transcripts
NM_001122979.2 NM_001170631.1 NM_032029.4 XM_011510051.2 XM_011510052.2 XM_017002442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7324286 c.797A>G p.*266* Chr.1: 206958651-206958651 on Build GRCh38
7324285 c.1599A>G p.L533L Chr.1: 206958651-206958651 on Build GRCh38
4027569 c.1622A>G p.E541G Chr.1: 206958628-206958628 on Build GRCh38
6218920 c.1619T>A p.L540Q Chr.1: 206958631-206958631 on Build GRCh38
7415315 c.1729C>A p.P577T Chr.1: 206958521-206958521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749562507 Chr.1: 206958559-206958559 on Build GRCh38
rs775640081 Chr.1: 206958483-206958483 on Build GRCh38
rs758295007 Chr.1: 206958520-206958520 on Build GRCh38
rs890498867 Chr.1: 206958643-206958643 on Build GRCh38
rs768004868 Chr.1: 206958572-206958572 on Build GRCh38
rs41305098 Chr.1: 206958607-206958607 on Build GRCh38
rs773662170 Chr.1: 206958575-206958575 on Build GRCh38
rs752975338 Chr.1: 206958504-206958505 on Build GRCh38
rs926446142 Chr.1: 206958532-206958532 on Build GRCh38
rs761999991 Chr.1: 206958569-206958569 on Build GRCh38
rs184963036 Chr.1: 206958508-206958508 on Build GRCh38
rs762370128 Chr.1: 206958488-206958488 on Build GRCh38
rs756441771 Chr.1: 206958496-206958496 on Build GRCh38
rs772103789 Chr.1: 206958700-206958700 on Build GRCh38
rs374894378 Chr.1: 206958576-206958576 on Build GRCh38
rs775758372 Chr.1: 206958554-206958554 on Build GRCh38
rs1010295185 Chr.1: 206958665-206958665 on Build GRCh38
rs774967998 Chr.1: 206958487-206958487 on Build GRCh38
rs757911937 Chr.1: 206958632-206958632 on Build GRCh38
rs779283706 Chr.1: 206958686-206958686 on Build GRCh38
rs778898758 Chr.1: 206958678-206958678 on Build GRCh38
rs751499362 Chr.1: 206958661-206958661 on Build GRCh38
rs1048150987 Chr.1: 206958637-206958637 on Build GRCh38
rs781094828 Chr.1: 206958670-206958670 on Build GRCh38
rs753551566 Chr.1: 206958515-206958515 on Build GRCh38
rs778350262 Chr.1: 206958518-206958518 on Build GRCh38
rs752132778 Chr.1: 206958628-206958628 on Build GRCh38
rs759135181 Chr.1: 206958600-206958600 on Build GRCh38
rs748257958 Chr.1: 206958698-206958698 on Build GRCh38
rs180858792 Chr.1: 206958492-206958492 on Build GRCh38
rs777751108 Chr.1: 206958642-206958642 on Build GRCh38
rs754228871 Chr.1: 206958598-206958598 on Build GRCh38
rs747677778 Chr.1: 206958519-206958519 on Build GRCh38
rs773750766 Chr.1: 206958708-206958708 on Build GRCh38
rs942157612 Chr.1: 206958521-206958521 on Build GRCh38
rs372389528 Chr.1: 206958507-206958507 on Build GRCh38
rs777575031 Chr.1: 206958524-206958524 on Build GRCh38
rs555854947 Chr.1: 206958566-206958566 on Build GRCh38
rs61729353 Chr.1: 206958533-206958533 on Build GRCh38
rs571268969 Chr.1: 206958484-206958484 on Build GRCh38
rs75773215 Chr.1: 206958706-206958706 on Build GRCh38
rs760345953 Chr.1: 206958514-206958514 on Build GRCh38
rs372153877 Chr.1: 206958561-206958561 on Build GRCh38
rs899051250 Chr.1: 206958595-206958595 on Build GRCh38
rs929244843 Chr.1: 206958596-206958596 on Build GRCh38
rs745680811 Chr.1: 206958677-206958677 on Build GRCh38
rs757044979 Chr.1: 206958668-206958668 on Build GRCh38
rs770689295 Chr.1: 206958536-206958536 on Build GRCh38
rs575799213 Chr.1: 206958577-206958577 on Build GRCh38

View Full Product Details