Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 20769865-20770138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOCAD
Entrez Gene ID
54914
Gene Name
focadhesin
Gene Aliases
KIAA1797
UniGene
Hs.136247
Gene Chromosome Location
Chr.9: 20658309-20995955 on Build GRCh38
Transcripts
NM_017794.4 XM_005251494.3 XM_011517945.2 XM_017014852.1 XM_017014853.1 XM_017014854.1 XM_017014855.1 XM_017014856.1 XM_017014857.1 XM_017014858.1 XM_017014859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5421645 c.700G>A p.V234I Chr.9: 20770032-20770032 on Build GRCh38
455756 c.793C>T p.L265F Chr.9: 20770125-20770125 on Build GRCh38
5899268 c.767G>T p.R256L Chr.9: 20770099-20770099 on Build GRCh38
1636132 c.729G>A p.A243A Chr.9: 20770061-20770061 on Build GRCh38
5643296 c.732G>A p.M244I Chr.9: 20770064-20770064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939319632 Chr.9: 20769901-20769901 on Build GRCh38
rs750911901 Chr.9: 20770103-20770103 on Build GRCh38
rs757622242 Chr.9: 20770040-20770040 on Build GRCh38
rs183007490 Chr.9: 20770104-20770104 on Build GRCh38
rs771891738 Chr.9: 20770070-20770070 on Build GRCh38
rs546587667 Chr.9: 20770015-20770015 on Build GRCh38
rs1055449951 Chr.9: 20769921-20769921 on Build GRCh38
rs544176915 Chr.9: 20769927-20769927 on Build GRCh38
rs760602881 Chr.9: 20770080-20770080 on Build GRCh38
rs376370384 Chr.9: 20770018-20770018 on Build GRCh38
rs752435985 Chr.9: 20769955-20769955 on Build GRCh38
rs10441706 Chr.9: 20770032-20770032 on Build GRCh38
rs537981944 Chr.9: 20770107-20770107 on Build GRCh38
rs933814958 Chr.9: 20769900-20769900 on Build GRCh38
rs776076815 Chr.9: 20770083-20770083 on Build GRCh38
rs7860490 Chr.9: 20769988-20769988 on Build GRCh38
rs1038431345 Chr.9: 20769992-20769992 on Build GRCh38
rs563975023 Chr.9: 20769970-20769970 on Build GRCh38
rs1029750963 Chr.9: 20770038-20770038 on Build GRCh38
rs1050967507 Chr.9: 20769908-20769908 on Build GRCh38
rs773024068 Chr.9: 20770079-20770079 on Build GRCh38
rs762151102 Chr.9: 20770098-20770098 on Build GRCh38
rs770449668 Chr.9: 20770082-20770082 on Build GRCh38
rs774242242 Chr.9: 20769997-20769997 on Build GRCh38
rs753502682 Chr.9: 20770017-20770017 on Build GRCh38
rs779170000 Chr.9: 20770061-20770061 on Build GRCh38
rs761635300 Chr.9: 20770003-20770003 on Build GRCh38
rs530555906 Chr.9: 20769923-20769923 on Build GRCh38
rs549463779 Chr.9: 20770058-20770058 on Build GRCh38
rs908451816 Chr.9: 20769926-20769926 on Build GRCh38
rs773183965 Chr.9: 20770010-20770010 on Build GRCh38
rs141558162 Chr.9: 20770060-20770060 on Build GRCh38
rs781607422 Chr.9: 20770041-20770041 on Build GRCh38
rs749458393 Chr.9: 20769987-20769987 on Build GRCh38
rs895171567 Chr.9: 20769956-20769956 on Build GRCh38
rs199678370 Chr.9: 20770090-20770090 on Build GRCh38
rs747948304 Chr.9: 20770062-20770062 on Build GRCh38
rs35122373 Chr.9: 20769901-20769902 on Build GRCh38
rs771950336 Chr.9: 20770009-20770009 on Build GRCh38
rs760166572 Chr.9: 20770011-20770011 on Build GRCh38
rs753793211 Chr.9: 20770099-20770099 on Build GRCh38
rs939972526 Chr.9: 20769972-20769972 on Build GRCh38
rs772825154 Chr.9: 20769997-20769998 on Build GRCh38
rs769982049 Chr.9: 20769983-20769983 on Build GRCh38
rs141572748 Chr.9: 20770051-20770051 on Build GRCh38
rs371251129 Chr.9: 20770019-20770019 on Build GRCh38
rs780228974 Chr.9: 20770055-20770055 on Build GRCh38
rs777207561 Chr.9: 20770091-20770091 on Build GRCh38
rs561463820 Chr.9: 20769913-20769913 on Build GRCh38

View Full Product Details