Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231336584-231336835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC8
Entrez Gene ID
149371
Gene Name
exocyst complex component 8
Gene Aliases
EXO84, Exo84p, SEC84
UniGene
Hs.356198
Gene Chromosome Location
Chr.1: 231332731-231337872 on Build GRCh38
Transcripts
NM_175876.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6192204 c.922G>A p.A308T Chr.1: 231336824-231336824 on Build GRCh38
7411352 c.1052G>A p.R351K Chr.1: 231336694-231336694 on Build GRCh38
6261798 c.1031T>A p.L344Q Chr.1: 231336715-231336715 on Build GRCh38
364944 c.943G>A p.D315N Chr.1: 231336803-231336803 on Build GRCh38
6244612 c.918C>A p.S306S Chr.1: 231336828-231336828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144793875 Chr.1: 231336791-231336791 on Build GRCh38
rs1039609498 Chr.1: 231336788-231336788 on Build GRCh38
rs746595093 Chr.1: 231336714-231336714 on Build GRCh38
rs756979233 Chr.1: 231336779-231336780 on Build GRCh38
rs776653782 Chr.1: 231336641-231336641 on Build GRCh38
rs781359798 Chr.1: 231336670-231336670 on Build GRCh38
rs367850043 Chr.1: 231336687-231336687 on Build GRCh38
rs779375116 Chr.1: 231336815-231336815 on Build GRCh38
rs770584042 Chr.1: 231336605-231336605 on Build GRCh38
rs773334020 Chr.1: 231336765-231336765 on Build GRCh38
rs745358364 Chr.1: 231336682-231336682 on Build GRCh38
rs111580847 Chr.1: 231336773-231336773 on Build GRCh38
rs771670194 Chr.1: 231336684-231336684 on Build GRCh38
rs745515473 Chr.1: 231336606-231336606 on Build GRCh38
rs148464625 Chr.1: 231336610-231336610 on Build GRCh38
rs752877756 Chr.1: 231336691-231336692 on Build GRCh38
rs374382054 Chr.1: 231336678-231336678 on Build GRCh38
rs143120742 Chr.1: 231336647-231336647 on Build GRCh38
rs768511878 Chr.1: 231336716-231336716 on Build GRCh38
rs766606141 Chr.1: 231336659-231336659 on Build GRCh38
rs761806541 Chr.1: 231336755-231336755 on Build GRCh38
rs778746897 Chr.1: 231336789-231336791 on Build GRCh38
rs377577608 Chr.1: 231336664-231336664 on Build GRCh38
rs750498559 Chr.1: 231336645-231336645 on Build GRCh38
rs760494680 Chr.1: 231336623-231336623 on Build GRCh38
rs745409431 Chr.1: 231336805-231336805 on Build GRCh38
rs1021390523 Chr.1: 231336625-231336625 on Build GRCh38
rs370183913 Chr.1: 231336672-231336672 on Build GRCh38
rs765149961 Chr.1: 231336762-231336762 on Build GRCh38
rs765305372 Chr.1: 231336643-231336643 on Build GRCh38
rs755095564 Chr.1: 231336781-231336781 on Build GRCh38
rs370713369 Chr.1: 231336814-231336814 on Build GRCh38
rs967491162 Chr.1: 231336626-231336626 on Build GRCh38
rs145329289 Chr.1: 231336774-231336774 on Build GRCh38
rs200964466 Chr.1: 231336660-231336660 on Build GRCh38
rs776341442 Chr.1: 231336752-231336752 on Build GRCh38
rs778402584 Chr.1: 231336801-231336801 on Build GRCh38
rs775175509 Chr.1: 231336686-231336686 on Build GRCh38
rs943359564 Chr.1: 231336775-231336775 on Build GRCh38
rs973941506 Chr.1: 231336725-231336725 on Build GRCh38
rs537536815 Chr.1: 231336795-231336795 on Build GRCh38
rs775206321 Chr.1: 231336618-231336618 on Build GRCh38
rs139980773 Chr.1: 231336633-231336633 on Build GRCh38
rs757955624 Chr.1: 231336807-231336807 on Build GRCh38
rs971193466 Chr.1: 231336707-231336707 on Build GRCh38
rs977121056 Chr.1: 231336632-231336632 on Build GRCh38
rs200538583 Chr.1: 231336768-231336768 on Build GRCh38

View Full Product Details