Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134886967-134887240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCN2
Entrez Gene ID
2220
Gene Name
ficolin 2
Gene Aliases
EBP-37, FCNL, P35, ficolin-2
UniGene
Hs.54517
Gene Chromosome Location
Chr.9: 134864146-134890506 on Build GRCh38
Transcripts
NM_004108.2 NM_015837.2 XM_006717015.3 XM_011518392.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3655498 c.763G>A p.G255R Chr.9: 134887236-134887236 on Build GRCh38
3736478 c.707C>T p.T236M Chr.9: 134887180-134887180 on Build GRCh38
3655497 c.702C>T p.S234S Chr.9: 134887175-134887175 on Build GRCh38
3375165 c.706A>G p.T236A Chr.9: 134887179-134887179 on Build GRCh38
5410687 c.764G>A p.G255E Chr.9: 134887237-134887237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs10125317 Chr.9: 134887037-134887037 on Build GRCh38
rs544869340 Chr.9: 134887079-134887079 on Build GRCh38
rs963470596 Chr.9: 134887172-134887172 on Build GRCh38
rs1034468582 Chr.9: 134887117-134887117 on Build GRCh38
rs377714485 Chr.9: 134887170-134887170 on Build GRCh38
rs900691665 Chr.9: 134887053-134887053 on Build GRCh38
rs778359530 Chr.9: 134886997-134886997 on Build GRCh38
rs7872508 Chr.9: 134887041-134887041 on Build GRCh38
rs373375538 Chr.9: 134887159-134887159 on Build GRCh38
rs1052478683 Chr.9: 134887101-134887101 on Build GRCh38
rs778609601 Chr.9: 134887061-134887061 on Build GRCh38
rs751231953 Chr.9: 134887185-134887185 on Build GRCh38
rs750421061 Chr.9: 134887165-134887165 on Build GRCh38
rs113125899 Chr.9: 134887166-134887166 on Build GRCh38
rs935707135 Chr.9: 134886994-134886994 on Build GRCh38
rs891424256 Chr.9: 134887007-134887007 on Build GRCh38
rs149631143 Chr.9: 134887202-134887202 on Build GRCh38
rs376615239 Chr.9: 134887134-134887134 on Build GRCh38
rs923512316 Chr.9: 134887194-134887194 on Build GRCh38
rs368792237 Chr.9: 134887143-134887143 on Build GRCh38
rs764017305 Chr.9: 134887181-134887181 on Build GRCh38
rs866705245 Chr.9: 134887177-134887177 on Build GRCh38
rs928308839 Chr.9: 134887002-134887002 on Build GRCh38
rs199873964 Chr.9: 134887132-134887132 on Build GRCh38
rs555522537 Chr.9: 134887030-134887030 on Build GRCh38
rs17549193 Chr.9: 134887180-134887180 on Build GRCh38
rs370925745 Chr.9: 134887142-134887142 on Build GRCh38
rs547815340 Chr.9: 134887193-134887193 on Build GRCh38
rs980450419 Chr.9: 134887001-134887001 on Build GRCh38
rs780414546 Chr.9: 134887212-134887212 on Build GRCh38
rs763259985 Chr.9: 134887106-134887106 on Build GRCh38
rs568247549 Chr.9: 134887174-134887174 on Build GRCh38
rs371307695 Chr.9: 134887186-134887186 on Build GRCh38
rs752737421 Chr.9: 134887177-134887177 on Build GRCh38
rs532083195 Chr.9: 134887099-134887099 on Build GRCh38
rs150323760 Chr.9: 134887069-134887069 on Build GRCh38
rs139535942 Chr.9: 134887188-134887188 on Build GRCh38
rs763249225 Chr.9: 134887119-134887119 on Build GRCh38
rs62573178 Chr.9: 134887004-134887004 on Build GRCh38
rs573645901 Chr.9: 134887036-134887036 on Build GRCh38
rs907085061 Chr.9: 134887098-134887098 on Build GRCh38
rs984868608 Chr.9: 134887171-134887171 on Build GRCh38
rs530625007 Chr.9: 134887112-134887112 on Build GRCh38

View Full Product Details