Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103328113-103328384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD6
Entrez Gene ID
8323
Gene Name
frizzled class receptor 6
Gene Aliases
FZ-6, FZ6, HFZ6, NDNC10
UniGene
Hs.591863
Gene Chromosome Location
Chr.8: 103298433-103332866 on Build GRCh38
Transcripts
NM_001164615.1 NM_001164616.1 NM_001317796.1 NM_003506.3 NR_133921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5873647 c.1480G>A p.V494I Chr.8: 103328355-103328355 on Build GRCh38
40120 c.1464C>T p.I488I Chr.8: 103328339-103328339 on Build GRCh38
3884266 c.1500C>T p.C500C Chr.8: 103328375-103328375 on Build GRCh38
1094465 c.1409G>A p.R470Q Chr.8: 103328284-103328284 on Build GRCh38
3643390 c.1419G>A p.L473L Chr.8: 103328294-103328294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894446091 Chr.8: 103328207-103328207 on Build GRCh38
rs368654295 Chr.8: 103328342-103328342 on Build GRCh38
rs149021436 Chr.8: 103328315-103328315 on Build GRCh38
rs758292286 Chr.8: 103328284-103328284 on Build GRCh38
rs780889553 Chr.8: 103328291-103328291 on Build GRCh38
rs1034638997 Chr.8: 103328190-103328190 on Build GRCh38
rs1034324229 Chr.8: 103328191-103328191 on Build GRCh38
rs376124026 Chr.8: 103328329-103328329 on Build GRCh38
rs781334683 Chr.8: 103328257-103328258 on Build GRCh38
rs777284498 Chr.8: 103328309-103328309 on Build GRCh38
rs747394243 Chr.8: 103328338-103328338 on Build GRCh38
rs147565570 Chr.8: 103328290-103328290 on Build GRCh38
rs751323317 Chr.8: 103328289-103328289 on Build GRCh38
rs904767868 Chr.8: 103328169-103328169 on Build GRCh38
rs371466170 Chr.8: 103328257-103328257 on Build GRCh38
rs756470480 Chr.8: 103328315-103328318 on Build GRCh38
rs780440018 Chr.8: 103328320-103328326 on Build GRCh38
rs762104663 Chr.8: 103328345-103328345 on Build GRCh38
rs536810006 Chr.8: 103328141-103328141 on Build GRCh38
rs773197811 Chr.8: 103328354-103328354 on Build GRCh38
rs768721252 Chr.8: 103328305-103328305 on Build GRCh38
rs555570719 Chr.8: 103328198-103328198 on Build GRCh38
rs750436184 Chr.8: 103328283-103328283 on Build GRCh38
rs752314851 Chr.8: 103328292-103328292 on Build GRCh38
rs780555971 Chr.8: 103328331-103328331 on Build GRCh38
rs747028715 Chr.8: 103328212-103328212 on Build GRCh38
rs755666655 Chr.8: 103328303-103328303 on Build GRCh38
rs1012295181 Chr.8: 103328228-103328228 on Build GRCh38
rs761036446 Chr.8: 103328249-103328249 on Build GRCh38
rs770093420 Chr.8: 103328353-103328353 on Build GRCh38
rs764239004 Chr.8: 103328256-103328256 on Build GRCh38
rs766301901 Chr.8: 103328288-103328288 on Build GRCh38
rs772578075 Chr.8: 103328232-103328232 on Build GRCh38
rs573632793 Chr.8: 103328246-103328246 on Build GRCh38
rs1042392656 Chr.8: 103328159-103328159 on Build GRCh38
rs769142373 Chr.8: 103328340-103328340 on Build GRCh38
rs868392863 Chr.8: 103328265-103328265 on Build GRCh38
rs371604427 Chr.8: 103328234-103328234 on Build GRCh38
rs750706481 Chr.8: 103328270-103328270 on Build GRCh38
rs765376606 Chr.8: 103328274-103328274 on Build GRCh38
rs113688012 Chr.8: 103328356-103328356 on Build GRCh38

View Full Product Details