Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86478866-86479127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN1
Entrez Gene ID
51115
Gene Name
regulator of microtubule dynamics 1
Gene Aliases
CGI-90, FAM82B, RMD-1, RMD1
UniGene
Hs.145386
Gene Chromosome Location
Chr.8: 86467116-86514348 on Build GRCh38
Transcripts
NM_001286707.1 NM_001286719.1 NM_001317807.1 NM_016033.2 XM_005250944.3 XM_005250947.3 XM_005250948.3 XM_006716573.3 XM_011517090.2 XM_011517091.2 XM_011517092.2 XM_011517093.2 XM_011517094.2 XM_011517095.2 XM_011517096.2 XM_011517097.2 XM_011517098.2 XM_017013518.1 XM_017013519.1 XM_017013520.1 XM_017013521.1 XM_017013522.1 XM_017013523.1 XM_017013524.1 XM_017013525.1 XM_017013526.1 XM_017013527.1 XM_017013528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4151913 c.642-8C>A p.? Chr.8: 86479018-86479018 on Build GRCh38
6193732 c.671A>G p.Y224C Chr.8: 86478981-86478981 on Build GRCh38
3663911 c.711T>A p.P237P Chr.8: 86478941-86478941 on Build GRCh38
6767020 c.680_682delGAA p.R227delR Chr.8: 86478970-86478972 on Build GRCh38
3901870 c.677G>A p.R226K Chr.8: 86478975-86478975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753751486 Chr.8: 86478892-86478892 on Build GRCh38
rs528407110 Chr.8: 86478893-86478893 on Build GRCh38
rs1020895983 Chr.8: 86478900-86478900 on Build GRCh38
rs775749961 Chr.8: 86478998-86478998 on Build GRCh38
rs769024706 Chr.8: 86478995-86478995 on Build GRCh38
rs747778858 Chr.8: 86479091-86479091 on Build GRCh38
rs757855099 Chr.8: 86478930-86478930 on Build GRCh38
rs551351368 Chr.8: 86479005-86479005 on Build GRCh38
rs771191809 Chr.8: 86478970-86478972 on Build GRCh38
rs773151714 Chr.8: 86479036-86479036 on Build GRCh38
rs761598002 Chr.8: 86479023-86479023 on Build GRCh38
rs779577121 Chr.8: 86478988-86478988 on Build GRCh38
rs903339056 Chr.8: 86478934-86478934 on Build GRCh38
rs757772860 Chr.8: 86478958-86478958 on Build GRCh38
rs764719202 Chr.8: 86478906-86478906 on Build GRCh38
rs760054026 Chr.8: 86479062-86479067 on Build GRCh38
rs756481931 Chr.8: 86478953-86478953 on Build GRCh38
rs749692012 Chr.8: 86478891-86478891 on Build GRCh38
rs974348714 Chr.8: 86479037-86479037 on Build GRCh38
rs751212714 Chr.8: 86479057-86479057 on Build GRCh38
rs749864997 Chr.8: 86478918-86478918 on Build GRCh38
rs765097095 Chr.8: 86479034-86479034 on Build GRCh38
rs1028652638 Chr.8: 86479049-86479049 on Build GRCh38
rs778341673 Chr.8: 86478956-86478956 on Build GRCh38
rs537242354 Chr.8: 86479085-86479085 on Build GRCh38
rs765859451 Chr.8: 86479051-86479051 on Build GRCh38
rs766019117 Chr.8: 86478949-86478949 on Build GRCh38
rs959627198 Chr.8: 86479038-86479038 on Build GRCh38
rs747307921 Chr.8: 86478903-86478905 on Build GRCh38
rs751214829 Chr.8: 86478951-86478951 on Build GRCh38
rs377525078 Chr.8: 86479011-86479011 on Build GRCh38
rs777206181 Chr.8: 86479057-86479061 on Build GRCh38
rs571449684 Chr.8: 86479010-86479010 on Build GRCh38
rs1020695287 Chr.8: 86478989-86478989 on Build GRCh38
rs745911569 Chr.8: 86478994-86478994 on Build GRCh38
rs773879012 Chr.8: 86479044-86479044 on Build GRCh38
rs777830408 Chr.8: 86478899-86478902 on Build GRCh38
rs762497589 Chr.8: 86479040-86479040 on Build GRCh38

View Full Product Details