Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112781407-112781680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1A
Entrez Gene ID
3552
Gene Name
interleukin 1 alpha
Gene Aliases
IL-1A, IL1, IL1-ALPHA, IL1F1
UniGene
Hs.1722
Gene Chromosome Location
Chr.2: 112773915-112785398 on Build GRCh38
Transcripts
NM_000575.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1210532 c.319+2T>C p.? Chr.2: 112781602-112781602 on Build GRCh38
6211043 c.312A>G p.S104S Chr.2: 112781611-112781611 on Build GRCh38
7087510 c.276C>T p.I92I Chr.2: 112781647-112781647 on Build GRCh38
5588041 c.272C>T p.S91F Chr.2: 112781651-112781651 on Build GRCh38
7372036 c.260G>C p.S87T Chr.2: 112781663-112781663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866459720 Chr.2: 112781607-112781607 on Build GRCh38
rs781599647 Chr.2: 112781601-112781601 on Build GRCh38
rs768230643 Chr.2: 112781628-112781628 on Build GRCh38
rs527883350 Chr.2: 112781477-112781477 on Build GRCh38
rs373905522 Chr.2: 112781520-112781520 on Build GRCh38
rs547951783 Chr.2: 112781500-112781500 on Build GRCh38
rs942314122 Chr.2: 112781473-112781473 on Build GRCh38
rs375431463 Chr.2: 112781653-112781653 on Build GRCh38
rs1039269324 Chr.2: 112781476-112781476 on Build GRCh38
rs149073466 Chr.2: 112781524-112781524 on Build GRCh38
rs143855190 Chr.2: 112781622-112781622 on Build GRCh38
rs20540 Chr.2: 112781647-112781647 on Build GRCh38
rs1052971323 Chr.2: 112781534-112781534 on Build GRCh38
rs1011929258 Chr.2: 112781588-112781588 on Build GRCh38
rs868583894 Chr.2: 112781651-112781651 on Build GRCh38
rs775579895 Chr.2: 112781483-112781483 on Build GRCh38
rs909377052 Chr.2: 112781436-112781436 on Build GRCh38
rs140186374 Chr.2: 112781615-112781615 on Build GRCh38
rs965982868 Chr.2: 112781512-112781512 on Build GRCh38
rs763121818 Chr.2: 112781584-112781584 on Build GRCh38
rs756490345 Chr.2: 112781610-112781610 on Build GRCh38
rs779488667 Chr.2: 112781623-112781623 on Build GRCh38
rs61538608 Chr.2: 112781631-112781631 on Build GRCh38
rs759016628 Chr.2: 112781505-112781505 on Build GRCh38
rs751811065 Chr.2: 112781595-112781595 on Build GRCh38
rs769546045 Chr.2: 112781650-112781650 on Build GRCh38
rs79919093 Chr.2: 112781618-112781618 on Build GRCh38
rs142134831 Chr.2: 112781655-112781655 on Build GRCh38
rs565286131 Chr.2: 112781451-112781451 on Build GRCh38
rs764688691 Chr.2: 112781600-112781600 on Build GRCh38
rs920947041 Chr.2: 112781499-112781499 on Build GRCh38
rs753043970 Chr.2: 112781603-112781603 on Build GRCh38
rs776161770 Chr.2: 112781645-112781645 on Build GRCh38
rs370435712 Chr.2: 112781582-112781582 on Build GRCh38
rs746553558 Chr.2: 112781624-112781624 on Build GRCh38

View Full Product Details