Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 39103855-39104120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP3
Entrez Gene ID
79937
Gene Name
contactin associated protein-like 3
Gene Aliases
CASPR3, CNTNAP3A
UniGene
Hs.128474
Gene Chromosome Location
Chr.9: 39072767-39288303 on Build GRCh38
Transcripts
NM_033655.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6299489 c.2109C>T p.F703F Chr.9: 39103907-39103907 on Build GRCh38
6299488 c.2373C>T p.F791F Chr.9: 39103907-39103907 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759726807 Chr.9: 39103885-39103885 on Build GRCh38
rs762809370 Chr.9: 39103913-39103914 on Build GRCh38
rs770780626 Chr.9: 39103881-39103881 on Build GRCh38
rs3119738 Chr.9: 39104021-39104021 on Build GRCh38
rs548180889 Chr.9: 39104069-39104069 on Build GRCh38
rs1023134492 Chr.9: 39104070-39104070 on Build GRCh38
rs532067833 Chr.9: 39103887-39103887 on Build GRCh38
rs375311457 Chr.9: 39103944-39103944 on Build GRCh38
rs570114583 Chr.9: 39104078-39104078 on Build GRCh38
rs756432693 Chr.9: 39104075-39104075 on Build GRCh38
rs1003137670 Chr.9: 39104087-39104087 on Build GRCh38
rs765733436 Chr.9: 39103935-39103935 on Build GRCh38
rs116002609 Chr.9: 39104088-39104088 on Build GRCh38
rs906078848 Chr.9: 39104083-39104084 on Build GRCh38
rs565706562 Chr.9: 39103937-39103937 on Build GRCh38
rs375854615 Chr.9: 39103882-39103882 on Build GRCh38
rs887945989 Chr.9: 39103956-39103956 on Build GRCh38
rs761179706 Chr.9: 39103925-39103925 on Build GRCh38
rs754608193 Chr.9: 39103942-39103942 on Build GRCh38
rs148916792 Chr.9: 39103907-39103907 on Build GRCh38
rs757689022 Chr.9: 39103934-39103934 on Build GRCh38

View Full Product Details