Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84150708-84150933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHITM
Entrez Gene ID
27069
Gene Name
growth hormone inducible transmembrane protein
Gene Aliases
DERP2, HSPC282, MICS1, My021, PTD010, TMBIM5
UniGene
Hs.352656
Gene Chromosome Location
Chr.10: 84139429-84153555 on Build GRCh38
Transcripts
NM_014394.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1253219 c.953C>A p.S318* Chr.10: 84150880-84150880 on Build GRCh38
7283341 c.805A>G p.T269A Chr.10: 84150732-84150732 on Build GRCh38
6670834 c.844G>A p.G282S Chr.10: 84150771-84150771 on Build GRCh38
6579284 c.850T>C p.L284L Chr.10: 84150777-84150777 on Build GRCh38
5596778 c.870C>T p.F290F Chr.10: 84150797-84150797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751397176 Chr.10: 84150784-84150784 on Build GRCh38
rs944418871 Chr.10: 84150867-84150867 on Build GRCh38
rs572795538 Chr.10: 84150829-84150829 on Build GRCh38
rs766979188 Chr.10: 84150896-84150897 on Build GRCh38
rs780778244 Chr.10: 84150797-84150797 on Build GRCh38
rs752105747 Chr.10: 84150858-84150858 on Build GRCh38
rs556751407 Chr.10: 84150758-84150758 on Build GRCh38
rs777286905 Chr.10: 84150906-84150906 on Build GRCh38
rs776322306 Chr.10: 84150743-84150743 on Build GRCh38
rs755359136 Chr.10: 84150870-84150870 on Build GRCh38
rs757193604 Chr.10: 84150811-84150811 on Build GRCh38
rs371326384 Chr.10: 84150770-84150770 on Build GRCh38
rs188887322 Chr.10: 84150827-84150827 on Build GRCh38
rs201822789 Chr.10: 84150762-84150762 on Build GRCh38
rs751915260 Chr.10: 84150791-84150791 on Build GRCh38
rs762040867 Chr.10: 84150771-84150771 on Build GRCh38
rs777005846 Chr.10: 84150832-84150832 on Build GRCh38
rs766831889 Chr.10: 84150856-84150856 on Build GRCh38
rs759314499 Chr.10: 84150828-84150828 on Build GRCh38
rs755335732 Chr.10: 84150794-84150794 on Build GRCh38
rs755724119 Chr.10: 84150888-84150888 on Build GRCh38
rs200385037 Chr.10: 84150750-84150750 on Build GRCh38
rs953479650 Chr.10: 84150764-84150764 on Build GRCh38
rs781482245 Chr.10: 84150874-84150874 on Build GRCh38
rs972242678 Chr.10: 84150815-84150815 on Build GRCh38
rs371764167 Chr.10: 84150826-84150826 on Build GRCh38
rs764273633 Chr.10: 84150766-84150766 on Build GRCh38
rs202027625 Chr.10: 84150881-84150881 on Build GRCh38
rs758930043 Chr.10: 84150779-84150779 on Build GRCh38
rs375934721 Chr.10: 84150780-84150780 on Build GRCh38
rs769310672 Chr.10: 84150805-84150805 on Build GRCh38
rs749254940 Chr.10: 84150822-84150822 on Build GRCh38
rs747699820 Chr.10: 84150745-84150745 on Build GRCh38
rs369560843 Chr.10: 84150837-84150837 on Build GRCh38
rs374633446 Chr.10: 84150734-84150734 on Build GRCh38
rs147214750 Chr.10: 84150839-84150839 on Build GRCh38
rs763064112 Chr.10: 84150848-84150848 on Build GRCh38
rs201409514 Chr.10: 84150765-84150765 on Build GRCh38
rs750969203 Chr.10: 84150775-84150775 on Build GRCh38
rs777198917 Chr.10: 84150818-84150818 on Build GRCh38
rs762823702 Chr.10: 84150757-84150757 on Build GRCh38
rs369199203 Chr.10: 84150803-84150803 on Build GRCh38

View Full Product Details