Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13090416-13090653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSG1
Entrez Gene ID
83445
Gene Name
germ cell associated 1
Gene Aliases
-
UniGene
Hs.240053
Gene Chromosome Location
Chr.12: 13083537-13103720 on Build GRCh38
Transcripts
NM_001080554.2 NM_001080555.2 NM_001206842.1 NM_001206843.1 NM_001206845.1 NM_031289.3 NM_153823.3 XM_005253493.2 XM_005253495.1 XM_011520858.1 XM_011520859.1 XM_011520860.1 XM_011520861.1 XM_011520862.1 XM_011520863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5374659 c.231C>T p.S77S Chr.12: 13090636-13090636 on Build GRCh38
6445741 c.333G>A p.W111* Chr.12: 13090534-13090534 on Build GRCh38
1360228 c.310C>T p.R104W Chr.12: 13090557-13090557 on Build GRCh38
4486587 c.306C>T p.S102S Chr.12: 13090561-13090561 on Build GRCh38
5735069 c.360A>T p.E120D Chr.12: 13090507-13090507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991970235 Chr.12: 13090525-13090525 on Build GRCh38
rs760020080 Chr.12: 13090597-13090597 on Build GRCh38
rs769734228 Chr.12: 13090569-13090569 on Build GRCh38
rs558492764 Chr.12: 13090622-13090622 on Build GRCh38
rs779122135 Chr.12: 13090568-13090568 on Build GRCh38
rs144983335 Chr.12: 13090602-13090602 on Build GRCh38
rs148269918 Chr.12: 13090450-13090450 on Build GRCh38
rs777168436 Chr.12: 13090593-13090593 on Build GRCh38
rs753886021 Chr.12: 13090615-13090615 on Build GRCh38
rs552505876 Chr.12: 13090493-13090493 on Build GRCh38
rs182359480 Chr.12: 13090567-13090567 on Build GRCh38
rs115405681 Chr.12: 13090589-13090589 on Build GRCh38
rs763575184 Chr.12: 13090506-13090506 on Build GRCh38
rs560828499 Chr.12: 13090456-13090456 on Build GRCh38
rs961587955 Chr.12: 13090483-13090483 on Build GRCh38
rs766256339 Chr.12: 13090610-13090610 on Build GRCh38
rs756934437 Chr.12: 13090545-13090545 on Build GRCh38
rs75744933 Chr.12: 13090582-13090582 on Build GRCh38
rs376150164 Chr.12: 13090491-13090491 on Build GRCh38
rs761174109 Chr.12: 13090600-13090600 on Build GRCh38
rs1010087107 Chr.12: 13090441-13090441 on Build GRCh38
rs199755415 Chr.12: 13090548-13090548 on Build GRCh38
rs140139872 Chr.12: 13090556-13090556 on Build GRCh38
rs763656506 Chr.12: 13090598-13090598 on Build GRCh38
rs868619740 Chr.12: 13090555-13090555 on Build GRCh38
rs750006525 Chr.12: 13090604-13090604 on Build GRCh38
rs371483265 Chr.12: 13090462-13090462 on Build GRCh38
rs373467560 Chr.12: 13090473-13090473 on Build GRCh38
rs773621859 Chr.12: 13090472-13090472 on Build GRCh38
rs11546332 Chr.12: 13090628-13090628 on Build GRCh38
rs369697259 Chr.12: 13090453-13090453 on Build GRCh38
rs149082366 Chr.12: 13090618-13090618 on Build GRCh38
rs968029931 Chr.12: 13090461-13090461 on Build GRCh38
rs773258832 Chr.12: 13090579-13090579 on Build GRCh38
rs772351734 Chr.12: 13090570-13090570 on Build GRCh38
rs570774377 Chr.12: 13090504-13090504 on Build GRCh38
rs536441717 Chr.12: 13090557-13090557 on Build GRCh38
rs371280503 Chr.12: 13090521-13090521 on Build GRCh38
rs773851434 Chr.12: 13090599-13090599 on Build GRCh38
rs528021155 Chr.12: 13090464-13090464 on Build GRCh38
rs760560382 Chr.12: 13090606-13090606 on Build GRCh38
rs531814550 Chr.12: 13090513-13090513 on Build GRCh38
rs144605733 Chr.12: 13090547-13090547 on Build GRCh38
rs1022413846 Chr.12: 13090442-13090442 on Build GRCh38
rs762662131 Chr.12: 13090589-13090590 on Build GRCh38
rs755249530 Chr.12: 13090552-13090552 on Build GRCh38

View Full Product Details