Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 121447586-121447843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA1
Entrez Gene ID
2697
Gene Name
gap junction protein alpha 1
Gene Aliases
AVSD3, CMDR, CX43, EKVP, GJAL, HLHS1, HSS, ODDD, PPKCA
UniGene
Hs.700699
Gene Chromosome Location
Chr.6: 121435577-121449744 on Build GRCh38
Transcripts
NM_000165.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3620063 c.747G>A p.A249A Chr.6: 121447594-121447594 on Build GRCh38
1440193 c.969G>A p.A323A Chr.6: 121447816-121447816 on Build GRCh38
3620064 c.803T>C p.F268S Chr.6: 121447650-121447650 on Build GRCh38
450422 c.950A>G p.Q317R Chr.6: 121447797-121447797 on Build GRCh38
6104744 c.942T>G p.S314R Chr.6: 121447789-121447789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562513101 Chr.6: 121447715-121447715 on Build GRCh38
rs766417328 Chr.6: 121447774-121447774 on Build GRCh38
rs778110855 Chr.6: 121447779-121447779 on Build GRCh38
rs138941844 Chr.6: 121447612-121447612 on Build GRCh38
rs753140658 Chr.6: 121447807-121447807 on Build GRCh38
rs774276262 Chr.6: 121447604-121447604 on Build GRCh38
rs748954821 Chr.6: 121447742-121447742 on Build GRCh38
rs758166001 Chr.6: 121447621-121447621 on Build GRCh38
rs775532447 Chr.6: 121447751-121447751 on Build GRCh38
rs55796648 Chr.6: 121447615-121447615 on Build GRCh38
rs777364271 Chr.6: 121447644-121447644 on Build GRCh38
rs762100241 Chr.6: 121447609-121447609 on Build GRCh38
rs747845039 Chr.6: 121447638-121447638 on Build GRCh38
rs758465154 Chr.6: 121447614-121447614 on Build GRCh38
rs976659652 Chr.6: 121447732-121447734 on Build GRCh38
rs149442143 Chr.6: 121447631-121447631 on Build GRCh38
rs143898957 Chr.6: 121447799-121447799 on Build GRCh38
rs569067227 Chr.6: 121447806-121447806 on Build GRCh38
rs781504444 Chr.6: 121447632-121447632 on Build GRCh38
rs765459582 Chr.6: 121447611-121447611 on Build GRCh38
rs756401045 Chr.6: 121447731-121447731 on Build GRCh38
rs778457698 Chr.6: 121447743-121447743 on Build GRCh38
rs768514685 Chr.6: 121447759-121447759 on Build GRCh38
rs773502253 Chr.6: 121447703-121447703 on Build GRCh38
rs370436837 Chr.6: 121447732-121447732 on Build GRCh38
rs759494167 Chr.6: 121447803-121447803 on Build GRCh38
rs772162102 Chr.6: 121447749-121447749 on Build GRCh38
rs746806096 Chr.6: 121447752-121447752 on Build GRCh38
rs555927773 Chr.6: 121447707-121447707 on Build GRCh38
rs745845216 Chr.6: 121447746-121447746 on Build GRCh38
rs17653265 Chr.6: 121447605-121447605 on Build GRCh38
rs751161110 Chr.6: 121447714-121447714 on Build GRCh38
rs760501433 Chr.6: 121447688-121447688 on Build GRCh38
rs769457947 Chr.6: 121447642-121447642 on Build GRCh38
rs764435531 Chr.6: 121447816-121447816 on Build GRCh38
rs777305726 Chr.6: 121447672-121447672 on Build GRCh38
rs773628965 Chr.6: 121447768-121447768 on Build GRCh38
rs756786635 Chr.6: 121447738-121447738 on Build GRCh38
rs767327649 Chr.6: 121447606-121447606 on Build GRCh38
rs753091320 Chr.6: 121447729-121447729 on Build GRCh38
rs759510089 Chr.6: 121447678-121447678 on Build GRCh38
rs774210184 Chr.6: 121447673-121447673 on Build GRCh38
rs752048581 Chr.6: 121447627-121447627 on Build GRCh38
rs756523929 Chr.6: 121447815-121447815 on Build GRCh38
rs767963064 Chr.6: 121447720-121447720 on Build GRCh38
rs67407537 Chr.6: 121447684-121447684 on Build GRCh38
rs753470523 Chr.6: 121447818-121447818 on Build GRCh38
rs376074787 Chr.6: 121447661-121447661 on Build GRCh38
rs763233834 Chr.6: 121447770-121447770 on Build GRCh38
rs765591338 Chr.6: 121447699-121447699 on Build GRCh38

View Full Product Details