Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72213835-72214098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDA
Entrez Gene ID
9615
Gene Name
guanine deaminase
Gene Aliases
CYPIN, GUANASE, NEDASIN
UniGene
Hs.494163
Gene Chromosome Location
Chr.9: 72114595-72256045 on Build GRCh38
Transcripts
NM_001242505.2 NM_001242506.2 NM_001242507.2 NM_004293.4 XM_005252317.1 XM_011519213.1 XM_011519214.1 XM_011519215.1 XM_011519216.1 XM_011519217.1 XM_011519218.2 XM_011519219.1 XM_011519220.1 XM_011519221.1 XM_011519222.1 XM_011519223.2 XM_017015333.1 XM_017015334.1 XM_017015335.1 XM_017015336.1 XM_017015337.1 XM_017015338.1 XM_017015339.1 XM_017015340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109968 c.488G>A p.R163Q Chr.9: 72213901-72213901 on Build GRCh38
3657993 c.578G>A p.R193K Chr.9: 72213991-72213991 on Build GRCh38
3657990 c.561G>A p.S187S Chr.9: 72213974-72213974 on Build GRCh38
3657987 c.560C>T p.S187L Chr.9: 72213973-72213973 on Build GRCh38
6048365 c.578+3delA p.? Chr.9: 72213994-72213994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752121579 Chr.9: 72213864-72213864 on Build GRCh38
rs17494333 Chr.9: 72214057-72214057 on Build GRCh38
rs1052807844 Chr.9: 72213900-72213900 on Build GRCh38
rs200156367 Chr.9: 72213940-72213940 on Build GRCh38
rs150730275 Chr.9: 72213974-72213974 on Build GRCh38
rs865863910 Chr.9: 72213957-72213957 on Build GRCh38
rs760095253 Chr.9: 72214035-72214035 on Build GRCh38
rs868568536 Chr.9: 72213947-72213947 on Build GRCh38
rs779287938 Chr.9: 72213934-72213934 on Build GRCh38
rs201577497 Chr.9: 72213918-72213918 on Build GRCh38
rs373720768 Chr.9: 72214015-72214015 on Build GRCh38
rs998511761 Chr.9: 72213888-72213888 on Build GRCh38
rs916176161 Chr.9: 72214073-72214073 on Build GRCh38
rs775128028 Chr.9: 72214034-72214034 on Build GRCh38
rs370708527 Chr.9: 72213999-72213999 on Build GRCh38
rs368217737 Chr.9: 72214045-72214045 on Build GRCh38
rs1027052641 Chr.9: 72213987-72213987 on Build GRCh38
rs760718998 Chr.9: 72213873-72213873 on Build GRCh38
rs747036622 Chr.9: 72213952-72213952 on Build GRCh38
rs368654878 Chr.9: 72213998-72213998 on Build GRCh38
rs1008781385 Chr.9: 72213911-72213911 on Build GRCh38
rs915375106 Chr.9: 72213978-72213978 on Build GRCh38
rs142048729 Chr.9: 72213908-72213908 on Build GRCh38
rs376451676 Chr.9: 72214013-72214013 on Build GRCh38
rs769274043 Chr.9: 72213968-72213968 on Build GRCh38
rs139088198 Chr.9: 72213982-72213982 on Build GRCh38
rs764092421 Chr.9: 72214043-72214043 on Build GRCh38
rs774026544 Chr.9: 72214007-72214007 on Build GRCh38
rs780222816 Chr.9: 72213950-72213950 on Build GRCh38

View Full Product Details