Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17579561-17579816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLGALT1
Entrez Gene ID
79709
Gene Name
collagen beta(1-O)galactosyltransferase 1
Gene Aliases
GLT25D1
UniGene
Hs.418795
Gene Chromosome Location
Chr.19: 17555594-17583162 on Build GRCh38
Transcripts
NM_024656.3 XM_005260080.4 XM_011528297.2 XM_011528298.2 XM_011528299.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4979753 c.1381G>A p.D461N Chr.19: 17579596-17579596 on Build GRCh38
5981489 c.1366G>A p.E456K Chr.19: 17579581-17579581 on Build GRCh38
5004671 c.1370G>A p.R457Q Chr.19: 17579585-17579585 on Build GRCh38
5056358 c.1373A>G p.E458G Chr.19: 17579588-17579588 on Build GRCh38
378255 c.1358G>A p.R453Q Chr.19: 17579573-17579573 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1034170387 Chr.19: 17579712-17579712 on Build GRCh38
rs182801383 Chr.19: 17579725-17579725 on Build GRCh38
rs961755376 Chr.19: 17579766-17579766 on Build GRCh38
rs1001311156 Chr.19: 17579709-17579709 on Build GRCh38
rs372493352 Chr.19: 17579621-17579621 on Build GRCh38
rs772059277 Chr.19: 17579657-17579657 on Build GRCh38
rs948615526 Chr.19: 17579738-17579738 on Build GRCh38
rs1044308765 Chr.19: 17579743-17579743 on Build GRCh38
rs1014964378 Chr.19: 17579763-17579763 on Build GRCh38
rs552521030 Chr.19: 17579602-17579602 on Build GRCh38
rs1045642750 Chr.19: 17579644-17579644 on Build GRCh38
rs374671467 Chr.19: 17579617-17579617 on Build GRCh38
rs145145339 Chr.19: 17579598-17579598 on Build GRCh38
rs532297516 Chr.19: 17579726-17579726 on Build GRCh38
rs907016454 Chr.19: 17579672-17579672 on Build GRCh38
rs201696003 Chr.19: 17579614-17579614 on Build GRCh38
rs995792111 Chr.19: 17579774-17579774 on Build GRCh38
rs770993973 Chr.19: 17579615-17579615 on Build GRCh38
rs777777218 Chr.19: 17579591-17579591 on Build GRCh38
rs1005975981 Chr.19: 17579749-17579749 on Build GRCh38
rs537054921 Chr.19: 17579777-17579777 on Build GRCh38
rs868214948 Chr.19: 17579739-17579739 on Build GRCh38
rs970270157 Chr.19: 17579794-17579794 on Build GRCh38
rs566133751 Chr.19: 17579674-17579674 on Build GRCh38
rs10407038 Chr.19: 17579773-17579773 on Build GRCh38
rs945917129 Chr.19: 17579632-17579632 on Build GRCh38
rs139673867 Chr.19: 17579585-17579585 on Build GRCh38
rs548707398 Chr.19: 17579755-17579755 on Build GRCh38
rs770482299 Chr.19: 17579616-17579616 on Build GRCh38
rs761711317 Chr.19: 17579655-17579655 on Build GRCh38
rs774422114 Chr.19: 17579627-17579627 on Build GRCh38
rs745798315 Chr.19: 17579581-17579581 on Build GRCh38
rs895754212 Chr.19: 17579677-17579677 on Build GRCh38

View Full Product Details