Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75541802-75542073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1
Entrez Gene ID
3092
Gene Name
huntingtin interacting protein 1
Gene Aliases
HIP-I, ILWEQ, SHON, SHONbeta, SHONgamma
UniGene
Hs.329266
Gene Chromosome Location
Chr.7: 75533298-75738976 on Build GRCh38
Transcripts
NM_001243198.2 NM_005338.6 XM_005250304.2 XM_005250305.2 XM_011516116.2 XM_017012099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5020678 c.2915C>T p.T972M Chr.7: 75541956-75541956 on Build GRCh38
1091752 c.2932C>T p.R978C Chr.7: 75541939-75541939 on Build GRCh38
3028500 c.2900A>T p.D967V Chr.7: 75541971-75541971 on Build GRCh38
4149345 c.2906C>G p.S969* Chr.7: 75541965-75541965 on Build GRCh38
3028499 c.2928C>A p.I976I Chr.7: 75541943-75541943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781980080 Chr.7: 75541938-75541938 on Build GRCh38
rs202215039 Chr.7: 75541901-75541901 on Build GRCh38
rs915312678 Chr.7: 75541886-75541886 on Build GRCh38
rs782523818 Chr.7: 75542017-75542017 on Build GRCh38
rs781918960 Chr.7: 75541946-75541946 on Build GRCh38
rs371815924 Chr.7: 75541939-75541939 on Build GRCh38
rs782415320 Chr.7: 75541911-75541911 on Build GRCh38
rs782299228 Chr.7: 75541890-75541890 on Build GRCh38
rs372182633 Chr.7: 75541892-75541892 on Build GRCh38
rs782504869 Chr.7: 75541974-75541974 on Build GRCh38
rs1039253151 Chr.7: 75541971-75541971 on Build GRCh38
rs782797327 Chr.7: 75541948-75541948 on Build GRCh38
rs782553872 Chr.7: 75541987-75541987 on Build GRCh38
rs782617238 Chr.7: 75541875-75541875 on Build GRCh38
rs376607958 Chr.7: 75541879-75541879 on Build GRCh38
rs75848794 Chr.7: 75541903-75541903 on Build GRCh38
rs782697345 Chr.7: 75541883-75541883 on Build GRCh38
rs782064470 Chr.7: 75541947-75541947 on Build GRCh38
rs782679366 Chr.7: 75542022-75542022 on Build GRCh38
rs375211454 Chr.7: 75541955-75541955 on Build GRCh38
rs781907345 Chr.7: 75541954-75541954 on Build GRCh38
rs781829115 Chr.7: 75541973-75541973 on Build GRCh38
rs781797433 Chr.7: 75542006-75542006 on Build GRCh38
rs139294983 Chr.7: 75541956-75541956 on Build GRCh38

View Full Product Details