Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96143832-96144077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP2
Entrez Gene ID
1844
Gene Name
dual specificity phosphatase 2
Gene Aliases
PAC-1, PAC1
UniGene
Hs.1183
Gene Chromosome Location
Chr.2: 96143166-96145468 on Build GRCh38
Transcripts
NM_004418.3 XM_017003546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5397589 c.844G>A p.E282K Chr.2: 96143924-96143924 on Build GRCh38
5714814 c.917A>T p.Q306L Chr.2: 96143851-96143851 on Build GRCh38
7095586 c.868C>T p.R290C Chr.2: 96143900-96143900 on Build GRCh38
6205488 c.731-3C>T p.? Chr.2: 96144040-96144040 on Build GRCh38
346682 c.930G>T p.Q310H Chr.2: 96143838-96143838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745791600 Chr.2: 96143867-96143867 on Build GRCh38
rs756203530 Chr.2: 96143983-96143983 on Build GRCh38
rs756683491 Chr.2: 96143924-96143924 on Build GRCh38
rs753630020 Chr.2: 96143897-96143897 on Build GRCh38
rs768360352 Chr.2: 96143890-96143890 on Build GRCh38
rs767865278 Chr.2: 96144052-96144052 on Build GRCh38
rs762740788 Chr.2: 96144048-96144048 on Build GRCh38
rs764902433 Chr.2: 96143970-96143970 on Build GRCh38
rs374111273 Chr.2: 96144012-96144012 on Build GRCh38
rs137991595 Chr.2: 96144015-96144015 on Build GRCh38
rs768849806 Chr.2: 96143932-96143932 on Build GRCh38
rs141345431 Chr.2: 96143938-96143938 on Build GRCh38
rs766312401 Chr.2: 96143961-96143961 on Build GRCh38
rs190055407 Chr.2: 96144025-96144025 on Build GRCh38
rs942386949 Chr.2: 96144013-96144013 on Build GRCh38
rs747889631 Chr.2: 96144020-96144020 on Build GRCh38
rs775141232 Chr.2: 96144044-96144044 on Build GRCh38
rs373647887 Chr.2: 96144009-96144009 on Build GRCh38
rs771621313 Chr.2: 96144021-96144021 on Build GRCh38
rs370789460 Chr.2: 96143975-96143975 on Build GRCh38
rs982317182 Chr.2: 96143881-96143881 on Build GRCh38
rs770902155 Chr.2: 96143992-96143992 on Build GRCh38
rs747722629 Chr.2: 96143941-96143941 on Build GRCh38
rs749118400 Chr.2: 96143890-96143891 on Build GRCh38
rs759397760 Chr.2: 96143969-96143969 on Build GRCh38
rs569902661 Chr.2: 96144027-96144027 on Build GRCh38
rs760865185 Chr.2: 96144057-96144057 on Build GRCh38
rs150034773 Chr.2: 96143927-96143927 on Build GRCh38
rs888536166 Chr.2: 96143895-96143895 on Build GRCh38
rs535465705 Chr.2: 96144029-96144029 on Build GRCh38
rs182119570 Chr.2: 96144026-96144026 on Build GRCh38
rs750624388 Chr.2: 96144054-96144054 on Build GRCh38
rs777343774 Chr.2: 96144024-96144024 on Build GRCh38
rs771931357 Chr.2: 96143899-96143899 on Build GRCh38
rs201899099 Chr.2: 96143909-96143909 on Build GRCh38
rs1011014740 Chr.2: 96144014-96144014 on Build GRCh38
rs760676017 Chr.2: 96143952-96143952 on Build GRCh38
rs377434804 Chr.2: 96143892-96143892 on Build GRCh38
rs138796808 Chr.2: 96143942-96143942 on Build GRCh38
rs373082796 Chr.2: 96143891-96143891 on Build GRCh38
rs372357109 Chr.2: 96143981-96143981 on Build GRCh38
rs141261455 Chr.2: 96143991-96143991 on Build GRCh38
rs755394685 Chr.2: 96144047-96144047 on Build GRCh38
rs866401448 Chr.2: 96143960-96143960 on Build GRCh38
rs749013732 Chr.2: 96143880-96143880 on Build GRCh38
rs147952657 Chr.2: 96143910-96143910 on Build GRCh38
rs776496987 Chr.2: 96143965-96143965 on Build GRCh38
rs746339897 Chr.2: 96143852-96143852 on Build GRCh38
rs561327206 Chr.2: 96143976-96143976 on Build GRCh38
rs369320464 Chr.2: 96143893-96143893 on Build GRCh38
rs570661321 Chr.2: 96143982-96143982 on Build GRCh38
rs144927730 Chr.2: 96143990-96143990 on Build GRCh38
rs757400657 Chr.2: 96143980-96143980 on Build GRCh38
rs977626489 Chr.2: 96143939-96143939 on Build GRCh38
rs780640891 Chr.2: 96143925-96143925 on Build GRCh38
rs749713427 Chr.2: 96143992-96143992 on Build GRCh38
rs545914740 Chr.2: 96143853-96143853 on Build GRCh38
rs184429608 Chr.2: 96143949-96143949 on Build GRCh38
rs760580274 Chr.2: 96143896-96143896 on Build GRCh38

View Full Product Details