Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174977442-174977705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM29
Entrez Gene ID
11086
Gene Name
ADAM metallopeptidase domain 29
Gene Aliases
CT73, svph1
UniGene
Hs.126838
Gene Chromosome Location
Chr.4: 174918358-174978180 on Build GRCh38
Transcripts
NM_001130703.1 NM_001130704.1 NM_001130705.1 NM_001278125.1 NM_001278126.1 NM_001278127.1 NM_014269.4 XM_011531556.1 XM_011531557.1 XM_011531559.1 XM_011531560.1 XM_011531561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6166993 c.1956C>A p.N652K Chr.4: 174977481-174977481 on Build GRCh38
481073 c.2128G>T p.V710F Chr.4: 174977653-174977653 on Build GRCh38
5689239 c.2122C>A p.Q708K Chr.4: 174977647-174977647 on Build GRCh38
6031183 c.2095A>T p.K699* Chr.4: 174977620-174977620 on Build GRCh38
5673721 c.2154G>A p.E718E Chr.4: 174977679-174977679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200852076 Chr.4: 174977593-174977595 on Build GRCh38
rs776159950 Chr.4: 174977596-174977600 on Build GRCh38
rs115171927 Chr.4: 174977559-174977559 on Build GRCh38
rs781501164 Chr.4: 174977481-174977481 on Build GRCh38
rs769292610 Chr.4: 174977529-174977529 on Build GRCh38
rs551435964 Chr.4: 174977665-174977665 on Build GRCh38
rs371841433 Chr.4: 174977515-174977515 on Build GRCh38
rs748696582 Chr.4: 174977499-174977499 on Build GRCh38
rs755382587 Chr.4: 174977473-174977473 on Build GRCh38
rs765280670 Chr.4: 174977557-174977557 on Build GRCh38
rs781647542 Chr.4: 174977581-174977581 on Build GRCh38
rs769393700 Chr.4: 174977504-174977504 on Build GRCh38
rs778385230 Chr.4: 174977650-174977650 on Build GRCh38
rs748743733 Chr.4: 174977612-174977612 on Build GRCh38
rs756740764 Chr.4: 174977590-174977590 on Build GRCh38
rs762890923 Chr.4: 174977681-174977681 on Build GRCh38
rs748931954 Chr.4: 174977523-174977523 on Build GRCh38
rs764369925 Chr.4: 174977677-174977679 on Build GRCh38
rs771926549 Chr.4: 174977547-174977547 on Build GRCh38
rs867831831 Chr.4: 174977544-174977544 on Build GRCh38
rs771733639 Chr.4: 174977663-174977663 on Build GRCh38
rs375497019 Chr.4: 174977676-174977676 on Build GRCh38
rs748102993 Chr.4: 174977642-174977642 on Build GRCh38
rs866176432 Chr.4: 174977541-174977541 on Build GRCh38
rs980229445 Chr.4: 174977673-174977673 on Build GRCh38
rs751802258 Chr.4: 174977472-174977472 on Build GRCh38
rs780455725 Chr.4: 174977466-174977466 on Build GRCh38
rs374967958 Chr.4: 174977502-174977502 on Build GRCh38
rs768648487 Chr.4: 174977674-174977674 on Build GRCh38
rs868811838 Chr.4: 174977474-174977474 on Build GRCh38
rs777415111 Chr.4: 174977602-174977602 on Build GRCh38
rs760647414 Chr.4: 174977553-174977553 on Build GRCh38
rs34732497 Chr.4: 174977620-174977620 on Build GRCh38
rs919925157 Chr.4: 174977570-174977570 on Build GRCh38
rs759596528 Chr.4: 174977629-174977629 on Build GRCh38
rs369083061 Chr.4: 174977550-174977550 on Build GRCh38
rs1008213796 Chr.4: 174977566-174977566 on Build GRCh38
rs141043508 Chr.4: 174977660-174977660 on Build GRCh38
rs770491554 Chr.4: 174977631-174977631 on Build GRCh38
rs776560687 Chr.4: 174977677-174977677 on Build GRCh38
rs866772953 Chr.4: 174977527-174977527 on Build GRCh38
rs759197670 Chr.4: 174977545-174977545 on Build GRCh38
rs751828290 Chr.4: 174977573-174977573 on Build GRCh38
rs780508389 Chr.4: 174977580-174977580 on Build GRCh38
rs750561912 Chr.4: 174977558-174977558 on Build GRCh38
rs560731315 Chr.4: 174977532-174977532 on Build GRCh38
rs563089840 Chr.4: 174977567-174977567 on Build GRCh38
rs760086310 Chr.4: 174977592-174977598 on Build GRCh38
rs772918991 Chr.4: 174977581-174977581 on Build GRCh38
rs746937720 Chr.4: 174977669-174977669 on Build GRCh38
rs753230754 Chr.4: 174977587-174977587 on Build GRCh38
rs201220428 Chr.4: 174977534-174977534 on Build GRCh38
rs866146754 Chr.4: 174977586-174977586 on Build GRCh38

View Full Product Details