Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12930134-12930367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARSA
Entrez Gene ID
2193
Gene Name
phenylalanyl-tRNA synthetase alpha subunit
Gene Aliases
CML33, FARSL, FARSLA, FRSA, PheHA
UniGene
Hs.23111
Gene Chromosome Location
Chr.19: 12922470-12933744 on Build GRCh38
Transcripts
NM_004461.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5198310 c.437_438insG p.Q147fs*3 Chr.19: 12930288-12930289 on Build GRCh38
4993959 c.403G>T p.E135* Chr.19: 12930323-12930323 on Build GRCh38
3528931 c.432G>A p.R144R Chr.19: 12930294-12930294 on Build GRCh38
3528932 c.400G>A p.D134N Chr.19: 12930326-12930326 on Build GRCh38
141548 c.430C>T p.R144W Chr.19: 12930296-12930296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371047882 Chr.19: 12930313-12930313 on Build GRCh38
rs748920575 Chr.19: 12930249-12930249 on Build GRCh38
rs150666819 Chr.19: 12930237-12930237 on Build GRCh38
rs144968946 Chr.19: 12930293-12930293 on Build GRCh38
rs113802496 Chr.19: 12930220-12930220 on Build GRCh38
rs1052327722 Chr.19: 12930163-12930163 on Build GRCh38
rs767858371 Chr.19: 12930247-12930247 on Build GRCh38
rs367913864 Chr.19: 12930263-12930263 on Build GRCh38
rs200098140 Chr.19: 12930292-12930292 on Build GRCh38
rs745402939 Chr.19: 12930322-12930322 on Build GRCh38
rs61737507 Chr.19: 12930317-12930317 on Build GRCh38
rs374285875 Chr.19: 12930336-12930336 on Build GRCh38
rs112863124 Chr.19: 12930221-12930221 on Build GRCh38
rs997945714 Chr.19: 12930158-12930168 on Build GRCh38
rs372267237 Chr.19: 12930338-12930338 on Build GRCh38
rs567765980 Chr.19: 12930311-12930311 on Build GRCh38
rs778484025 Chr.19: 12930261-12930261 on Build GRCh38
rs1043309876 Chr.19: 12930306-12930306 on Build GRCh38
rs766543627 Chr.19: 12930209-12930209 on Build GRCh38
rs767523994 Chr.19: 12930308-12930308 on Build GRCh38
rs202157711 Chr.19: 12930295-12930295 on Build GRCh38
rs746493788 Chr.19: 12930289-12930289 on Build GRCh38
rs774492543 Chr.19: 12930298-12930298 on Build GRCh38
rs556276607 Chr.19: 12930314-12930314 on Build GRCh38
rs765577009 Chr.19: 12930184-12930184 on Build GRCh38
rs762022336 Chr.19: 12930181-12930181 on Build GRCh38
rs752561439 Chr.19: 12930227-12930227 on Build GRCh38
rs983270100 Chr.19: 12930248-12930248 on Build GRCh38
rs141921998 Chr.19: 12930234-12930234 on Build GRCh38
rs367716693 Chr.19: 12930191-12930191 on Build GRCh38
rs778375194 Chr.19: 12930321-12930321 on Build GRCh38
rs908655552 Chr.19: 12930289-12930289 on Build GRCh38
rs757804010 Chr.19: 12930337-12930337 on Build GRCh38
rs751507369 Chr.19: 12930211-12930211 on Build GRCh38
rs748549803 Chr.19: 12930290-12930290 on Build GRCh38
rs182269315 Chr.19: 12930180-12930180 on Build GRCh38
rs374032555 Chr.19: 12930204-12930204 on Build GRCh38
rs532299148 Chr.19: 12930172-12930172 on Build GRCh38
rs769216883 Chr.19: 12930276-12930276 on Build GRCh38
rs767301673 Chr.19: 12930217-12930217 on Build GRCh38
rs773779224 Chr.19: 12930255-12930255 on Build GRCh38
rs759486272 Chr.19: 12930300-12930300 on Build GRCh38
rs920485733 Chr.19: 12930207-12930207 on Build GRCh38
rs34795408 Chr.19: 12930240-12930240 on Build GRCh38
rs1039345146 Chr.19: 12930296-12930296 on Build GRCh38
rs770535855 Chr.19: 12930320-12930321 on Build GRCh38
rs199824396 Chr.19: 12930310-12930310 on Build GRCh38
rs776952004 Chr.19: 12930282-12930282 on Build GRCh38
rs777211508 Chr.19: 12930288-12930289 on Build GRCh38
rs149267610 Chr.19: 12930272-12930272 on Build GRCh38

View Full Product Details