Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13366596-13366853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
UniGene
Hs.65734
Gene Chromosome Location
Chr.11: 13276613-13387268 on Build GRCh38
Transcripts
NM_001030272.2 NM_001030273.2 NM_001178.5 NM_001297719.1 NM_001297722.1 NM_001297724.1 XM_006718233.3 XM_006718234.1 XM_011520105.2 XM_011520107.2 XM_011520108.1 XM_011520109.1 XM_011520110.1 XM_011520111.1 XM_011520112.2 XM_011520113.1 XM_017017738.1 XM_017017739.1 XM_017017740.1 XM_017017741.1 XM_017017742.1 XM_017017743.1 XM_017017744.1 XM_017017745.1 XM_017017746.1 XM_017017747.1 XM_017017748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3445485 c.593C>T p.P198L Chr.11: 13366705-13366705 on Build GRCh38
925134 c.661G>T p.D221Y Chr.11: 13366773-13366773 on Build GRCh38
5712564 c.636C>T p.D212D Chr.11: 13366748-13366748 on Build GRCh38
4663466 c.553-1G>T p.? Chr.11: 13366664-13366664 on Build GRCh38
5970091 c.568C>G p.Q190E Chr.11: 13366680-13366680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747837626 Chr.11: 13366652-13366652 on Build GRCh38
rs188144151 Chr.11: 13366749-13366749 on Build GRCh38
rs767607113 Chr.11: 13366700-13366700 on Build GRCh38
rs1016726277 Chr.11: 13366830-13366830 on Build GRCh38
rs373788070 Chr.11: 13366662-13366662 on Build GRCh38
rs779909829 Chr.11: 13366626-13366626 on Build GRCh38
rs372039252 Chr.11: 13366822-13366822 on Build GRCh38
rs773694219 Chr.11: 13366655-13366655 on Build GRCh38
rs749600335 Chr.11: 13366805-13366805 on Build GRCh38
rs772605211 Chr.11: 13366824-13366824 on Build GRCh38
rs771216468 Chr.11: 13366810-13366810 on Build GRCh38
rs144766524 Chr.11: 13366752-13366752 on Build GRCh38
rs769612972 Chr.11: 13366653-13366653 on Build GRCh38
rs367807656 Chr.11: 13366798-13366798 on Build GRCh38
rs780931196 Chr.11: 13366641-13366641 on Build GRCh38
rs780820836 Chr.11: 13366792-13366792 on Build GRCh38
rs1023286300 Chr.11: 13366790-13366790 on Build GRCh38
rs755819487 Chr.11: 13366799-13366799 on Build GRCh38
rs147248699 Chr.11: 13366679-13366679 on Build GRCh38
rs772113504 Chr.11: 13366789-13366790 on Build GRCh38
rs750216866 Chr.11: 13366766-13366766 on Build GRCh38
rs777487963 Chr.11: 13366627-13366627 on Build GRCh38
rs202059327 Chr.11: 13366630-13366630 on Build GRCh38
rs768052689 Chr.11: 13366632-13366632 on Build GRCh38
rs991155494 Chr.11: 13366697-13366697 on Build GRCh38
rs757010873 Chr.11: 13366764-13366764 on Build GRCh38
rs748423781 Chr.11: 13366644-13366645 on Build GRCh38
rs758190125 Chr.11: 13366751-13366751 on Build GRCh38
rs919099123 Chr.11: 13366809-13366809 on Build GRCh38
rs773319868 Chr.11: 13366820-13366823 on Build GRCh38
rs140803737 Chr.11: 13366706-13366706 on Build GRCh38
rs777637094 Chr.11: 13366802-13366802 on Build GRCh38
rs775524867 Chr.11: 13366831-13366831 on Build GRCh38
rs368050146 Chr.11: 13366765-13366765 on Build GRCh38
rs112626431 Chr.11: 13366699-13366699 on Build GRCh38
rs759604326 Chr.11: 13366698-13366698 on Build GRCh38
rs754500431 Chr.11: 13366791-13366791 on Build GRCh38
rs564117857 Chr.11: 13366813-13366813 on Build GRCh38
rs749676442 Chr.11: 13366659-13366659 on Build GRCh38
rs760855922 Chr.11: 13366748-13366748 on Build GRCh38

View Full Product Details