Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108854657-108854896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL7B
Entrez Gene ID
10880
Gene Name
actin like 7B
Gene Aliases
Tact1
UniGene
Hs.534390
Gene Chromosome Location
Chr.9: 108854588-108855995 on Build GRCh38
Transcripts
NM_006686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1103658 c.1244G>A p.C415Y Chr.9: 108854687-108854687 on Build GRCh38
5137312 c.1225G>A p.V409M Chr.9: 108854706-108854706 on Build GRCh38
2153331 c.1039A>G p.M347V Chr.9: 108854892-108854892 on Build GRCh38
4163196 c.1037C>G p.T346S Chr.9: 108854894-108854894 on Build GRCh38
273618 c.1061G>A p.R354H Chr.9: 108854870-108854870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370958419 Chr.9: 108854706-108854706 on Build GRCh38
rs770877963 Chr.9: 108854814-108854814 on Build GRCh38
rs972986619 Chr.9: 108854682-108854682 on Build GRCh38
rs1013829913 Chr.9: 108854839-108854839 on Build GRCh38
rs923154338 Chr.9: 108854695-108854695 on Build GRCh38
rs902317740 Chr.9: 108854800-108854800 on Build GRCh38
rs753089435 Chr.9: 108854871-108854871 on Build GRCh38
rs777449902 Chr.9: 108854875-108854875 on Build GRCh38
rs761854373 Chr.9: 108854804-108854805 on Build GRCh38
rs755365391 Chr.9: 108854771-108854771 on Build GRCh38
rs766908997 Chr.9: 108854857-108854857 on Build GRCh38
rs757396032 Chr.9: 108854755-108854755 on Build GRCh38
rs768336085 Chr.9: 108854698-108854698 on Build GRCh38
rs764376655 Chr.9: 108854742-108854742 on Build GRCh38
rs35433501 Chr.9: 108854727-108854728 on Build GRCh38
rs895187968 Chr.9: 108854732-108854732 on Build GRCh38
rs755522980 Chr.9: 108854868-108854868 on Build GRCh38
rs780478048 Chr.9: 108854764-108854764 on Build GRCh38
rs901751354 Chr.9: 108854860-108854860 on Build GRCh38
rs752064414 Chr.9: 108854859-108854859 on Build GRCh38
rs986490003 Chr.9: 108854754-108854754 on Build GRCh38
rs754175469 Chr.9: 108854752-108854752 on Build GRCh38
rs775436347 Chr.9: 108854830-108854830 on Build GRCh38
rs781470020 Chr.9: 108854869-108854869 on Build GRCh38
rs748769810 Chr.9: 108854809-108854809 on Build GRCh38
rs769443334 Chr.9: 108854794-108854794 on Build GRCh38
rs771048019 Chr.9: 108854677-108854677 on Build GRCh38
rs772668094 Chr.9: 108854795-108854795 on Build GRCh38
rs762057573 Chr.9: 108854835-108854835 on Build GRCh38
rs750564969 Chr.9: 108854760-108854760 on Build GRCh38
rs139044591 Chr.9: 108854841-108854841 on Build GRCh38
rs765347719 Chr.9: 108854756-108854756 on Build GRCh38
rs868000824 Chr.9: 108854711-108854711 on Build GRCh38
rs1042797226 Chr.9: 108854763-108854763 on Build GRCh38
rs776643262 Chr.9: 108854726-108854726 on Build GRCh38
rs759367053 Chr.9: 108854816-108854816 on Build GRCh38
rs762987318 Chr.9: 108854856-108854856 on Build GRCh38
rs765581337 Chr.9: 108854842-108854842 on Build GRCh38
rs747373089 Chr.9: 108854769-108854769 on Build GRCh38
rs759298090 Chr.9: 108854740-108854740 on Build GRCh38
rs772047400 Chr.9: 108854828-108854828 on Build GRCh38
rs759324444 Chr.9: 108854785-108854785 on Build GRCh38
rs780429221 Chr.9: 108854689-108854689 on Build GRCh38
rs551989310 Chr.9: 108854843-108854843 on Build GRCh38
rs774195611 Chr.9: 108854815-108854815 on Build GRCh38
rs768043823 Chr.9: 108854839-108854839 on Build GRCh38
rs774565163 Chr.9: 108854700-108854701 on Build GRCh38
rs368861770 Chr.9: 108854691-108854691 on Build GRCh38
rs150901542 Chr.9: 108854719-108854719 on Build GRCh38
rs924369296 Chr.9: 108854770-108854770 on Build GRCh38
rs769353389 Chr.9: 108854707-108854707 on Build GRCh38
rs939646974 Chr.9: 108854714-108854714 on Build GRCh38
rs946741542 Chr.9: 108854765-108854765 on Build GRCh38
rs758893098 Chr.9: 108854761-108854761 on Build GRCh38
rs762880009 Chr.9: 108854713-108854713 on Build GRCh38
rs776306463 Chr.9: 108854700-108854700 on Build GRCh38
rs755713651 Chr.9: 108854874-108854874 on Build GRCh38
rs375138636 Chr.9: 108854725-108854725 on Build GRCh38
rs375377607 Chr.9: 108854781-108854781 on Build GRCh38
rs370549060 Chr.9: 108854779-108854780 on Build GRCh38

View Full Product Details