Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1338773-1339030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP3
Entrez Gene ID
8938
Gene Name
BAI1 associated protein 3
Gene Aliases
BAP3
UniGene
Hs.458427
Gene Chromosome Location
Chr.16: 1333605-1349441 on Build GRCh38
Transcripts
NM_001199096.1 NM_001199097.1 NM_001199098.1 NM_001199099.1 NM_001286464.1 NM_003933.4 XM_011522728.1 XM_011522729.1 XM_011522730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6078381 c.295G>A p.E99K Chr.16: 1338960-1338960 on Build GRCh38
6838916 c.199C>G p.Q67E Chr.16: 1338969-1338969 on Build GRCh38
1323722 c.243C>T p.P81P Chr.16: 1338908-1338908 on Build GRCh38
5828335 c.138C>T p.P46P Chr.16: 1338908-1338908 on Build GRCh38
6555327 c.160G>T p.A54S Chr.16: 1338930-1338930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949746682 Chr.16: 1338845-1338845 on Build GRCh38
rs374032783 Chr.16: 1339013-1339013 on Build GRCh38
rs780040420 Chr.16: 1338900-1338900 on Build GRCh38
rs368521574 Chr.16: 1338863-1338863 on Build GRCh38
rs758611781 Chr.16: 1338984-1338984 on Build GRCh38
rs373387941 Chr.16: 1338986-1338986 on Build GRCh38
rs377745556 Chr.16: 1338977-1338977 on Build GRCh38
rs764919514 Chr.16: 1338883-1338883 on Build GRCh38
rs779116674 Chr.16: 1338974-1338974 on Build GRCh38
rs745373846 Chr.16: 1338992-1338992 on Build GRCh38
rs766863731 Chr.16: 1338970-1338970 on Build GRCh38
rs948424220 Chr.16: 1338856-1338856 on Build GRCh38
rs776052974 Chr.16: 1338924-1338924 on Build GRCh38
rs770795319 Chr.16: 1338987-1338987 on Build GRCh38
rs768403043 Chr.16: 1339000-1339000 on Build GRCh38
rs752970857 Chr.16: 1338979-1338979 on Build GRCh38
rs7197416 Chr.16: 1338887-1338887 on Build GRCh38
rs753934248 Chr.16: 1338906-1338906 on Build GRCh38
rs981477374 Chr.16: 1338852-1338852 on Build GRCh38
rs769487647 Chr.16: 1338994-1338994 on Build GRCh38
rs776197907 Chr.16: 1338871-1338871 on Build GRCh38
rs564004825 Chr.16: 1338850-1338850 on Build GRCh38
rs768061810 Chr.16: 1338953-1338953 on Build GRCh38
rs747945715 Chr.16: 1338910-1338910 on Build GRCh38
rs996875799 Chr.16: 1338957-1338957 on Build GRCh38
rs145548563 Chr.16: 1338958-1338958 on Build GRCh38
rs762682224 Chr.16: 1338945-1338945 on Build GRCh38
rs560040369 Chr.16: 1338818-1338818 on Build GRCh38
rs77633104 Chr.16: 1338908-1338908 on Build GRCh38
rs139445484 Chr.16: 1338909-1338909 on Build GRCh38
rs1025229622 Chr.16: 1338999-1338999 on Build GRCh38
rs775137248 Chr.16: 1338998-1338998 on Build GRCh38
rs956967460 Chr.16: 1338831-1338831 on Build GRCh38
rs889224499 Chr.16: 1338938-1338938 on Build GRCh38
rs777527471 Chr.16: 1338917-1338917 on Build GRCh38
rs771792597 Chr.16: 1338916-1338916 on Build GRCh38
rs377387295 Chr.16: 1339012-1339012 on Build GRCh38
rs149675788 Chr.16: 1338918-1338918 on Build GRCh38
rs144485372 Chr.16: 1338933-1338933 on Build GRCh38
rs774078922 Chr.16: 1338891-1338894 on Build GRCh38
rs138846734 Chr.16: 1338864-1338864 on Build GRCh38
rs756641821 Chr.16: 1338898-1338898 on Build GRCh38
rs370067004 Chr.16: 1338858-1338858 on Build GRCh38
rs956889411 Chr.16: 1338980-1338980 on Build GRCh38
rs544110355 Chr.16: 1338839-1338839 on Build GRCh38
rs773737203 Chr.16: 1339002-1339002 on Build GRCh38
rs34787656 Chr.16: 1338982-1338982 on Build GRCh38
rs768151751 Chr.16: 1338891-1338891 on Build GRCh38
rs751994231 Chr.16: 1338886-1338886 on Build GRCh38
rs753899566 Chr.16: 1338972-1338972 on Build GRCh38
rs775215356 Chr.16: 1338940-1338940 on Build GRCh38
rs369995845 Chr.16: 1338981-1338981 on Build GRCh38
rs139609870 Chr.16: 1338939-1338939 on Build GRCh38
rs888505134 Chr.16: 1338959-1338959 on Build GRCh38
rs770867768 Chr.16: 1338920-1338920 on Build GRCh38
rs540021066 Chr.16: 1338802-1338802 on Build GRCh38
rs925419091 Chr.16: 1338812-1338812 on Build GRCh38
rs761063151 Chr.16: 1339004-1339004 on Build GRCh38
rs747892050 Chr.16: 1338862-1338862 on Build GRCh38
rs755022889 Chr.16: 1338973-1338973 on Build GRCh38
rs534433844 Chr.16: 1338960-1338960 on Build GRCh38
rs774065039 Chr.16: 1338857-1338857 on Build GRCh38
rs993098494 Chr.16: 1338834-1338834 on Build GRCh38
rs759150181 Chr.16: 1338882-1338882 on Build GRCh38
rs751029676 Chr.16: 1338894-1338894 on Build GRCh38
rs960535965 Chr.16: 1338829-1338829 on Build GRCh38
rs988397739 Chr.16: 1338832-1338832 on Build GRCh38
rs765860992 Chr.16: 1338866-1338866 on Build GRCh38

View Full Product Details