Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7844945-7845198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP6
Entrez Gene ID
654
Gene Name
bone morphogenetic protein 6
Gene Aliases
VGR, VGR1
UniGene
Hs.285671
Gene Chromosome Location
Chr.6: 7726778-7881728 on Build GRCh38
Transcripts
NM_001718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1696910 c.679G>A p.E227K Chr.6: 7845154-7845154 on Build GRCh38
5005920 c.698G>A p.R233Q Chr.6: 7845173-7845173 on Build GRCh38
5820423 c.690T>C p.P230P Chr.6: 7845165-7845165 on Build GRCh38
6611970 c.672C>T p.Y224Y Chr.6: 7845147-7845147 on Build GRCh38
179931 c.692G>A p.R231H Chr.6: 7845167-7845167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020012332 Chr.6: 7845018-7845018 on Build GRCh38
rs566660170 Chr.6: 7845167-7845167 on Build GRCh38
rs756972558 Chr.6: 7845119-7845119 on Build GRCh38
rs371529658 Chr.6: 7845092-7845092 on Build GRCh38
rs1035865004 Chr.6: 7844982-7844982 on Build GRCh38
rs753718516 Chr.6: 7845109-7845109 on Build GRCh38
rs1029176272 Chr.6: 7845029-7845029 on Build GRCh38
rs987779464 Chr.6: 7845062-7845062 on Build GRCh38
rs757988937 Chr.6: 7845148-7845148 on Build GRCh38
rs866459610 Chr.6: 7845170-7845170 on Build GRCh38
rs898559597 Chr.6: 7845121-7845121 on Build GRCh38
rs549460318 Chr.6: 7844996-7844996 on Build GRCh38
rs74667885 Chr.6: 7845086-7845086 on Build GRCh38
rs778696459 Chr.6: 7845136-7845136 on Build GRCh38
rs746482197 Chr.6: 7845157-7845157 on Build GRCh38
rs571290365 Chr.6: 7845052-7845052 on Build GRCh38
rs375867934 Chr.6: 7845147-7845147 on Build GRCh38
rs779543759 Chr.6: 7845154-7845154 on Build GRCh38
rs200180054 Chr.6: 7845089-7845089 on Build GRCh38
rs1057185122 Chr.6: 7844981-7844981 on Build GRCh38
rs956201803 Chr.6: 7844993-7844993 on Build GRCh38
rs531617814 Chr.6: 7845012-7845012 on Build GRCh38
rs776460025 Chr.6: 7845122-7845124 on Build GRCh38
rs778119506 Chr.6: 7845172-7845172 on Build GRCh38
rs763891048 Chr.6: 7845095-7845095 on Build GRCh38
rs1001490136 Chr.6: 7844978-7844978 on Build GRCh38
rs989002561 Chr.6: 7845001-7845001 on Build GRCh38
rs961984376 Chr.6: 7845067-7845067 on Build GRCh38
rs201278326 Chr.6: 7845173-7845173 on Build GRCh38
rs911884415 Chr.6: 7845063-7845063 on Build GRCh38
rs539387066 Chr.6: 7845068-7845068 on Build GRCh38
rs769518399 Chr.6: 7845141-7845143 on Build GRCh38
rs747256757 Chr.6: 7844996-7844997 on Build GRCh38
rs775083136 Chr.6: 7845028-7845028 on Build GRCh38
rs759290904 Chr.6: 7845124-7845125 on Build GRCh38
rs76088603 Chr.6: 7845019-7845019 on Build GRCh38
rs267192 Chr.6: 7845011-7845011 on Build GRCh38
rs937254641 Chr.6: 7845149-7845149 on Build GRCh38

View Full Product Details