Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100088928-100089173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS6
Entrez Gene ID
10980
Gene Name
COP9 signalosome subunit 6
Gene Aliases
CSN6, MOV34-34KD
UniGene
Hs.15591
Gene Chromosome Location
Chr.7: 100088960-100092200 on Build GRCh38
Transcripts
NM_006833.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4553071 c.57G>A p.G19G Chr.7: 100089047-100089047 on Build GRCh38
4549685 c.47G>A p.G16E Chr.7: 100089037-100089037 on Build GRCh38
5351466 c.2_3insGGCGGC p.A10_T11insAA Chr.7: 100088992-100088993 on Build GRCh38
4149448 c.65T>G p.V22G Chr.7: 100089055-100089055 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369030775 Chr.7: 100089079-100089079 on Build GRCh38
rs1006414141 Chr.7: 100089016-100089016 on Build GRCh38
rs921550755 Chr.7: 100089071-100089071 on Build GRCh38
rs988198760 Chr.7: 100089028-100089028 on Build GRCh38
rs768124617 Chr.7: 100089005-100089006 on Build GRCh38
rs969202119 Chr.7: 100088958-100088958 on Build GRCh38
rs777015230 Chr.7: 100089006-100089006 on Build GRCh38
rs928285286 Chr.7: 100088983-100088984 on Build GRCh38
rs554226642 Chr.7: 100089005-100089005 on Build GRCh38
rs571882095 Chr.7: 100088992-100088993 on Build GRCh38
rs1031572242 Chr.7: 100089103-100089103 on Build GRCh38
rs754660745 Chr.7: 100088945-100088945 on Build GRCh38
rs940132923 Chr.7: 100089126-100089126 on Build GRCh38
rs975663347 Chr.7: 100089047-100089047 on Build GRCh38
rs965593420 Chr.7: 100089043-100089043 on Build GRCh38
rs536287932 Chr.7: 100088953-100088953 on Build GRCh38
rs769644738 Chr.7: 100088993-100088995 on Build GRCh38
rs576209115 Chr.7: 100089008-100089008 on Build GRCh38
rs537116137 Chr.7: 100089023-100089023 on Build GRCh38
rs577546796 Chr.7: 100089136-100089136 on Build GRCh38
rs990033588 Chr.7: 100089127-100089127 on Build GRCh38
rs1032502671 Chr.7: 100089022-100089022 on Build GRCh38
rs377525504 Chr.7: 100089099-100089099 on Build GRCh38
rs374716476 Chr.7: 100088995-100088995 on Build GRCh38
rs373350105 Chr.7: 100089090-100089090 on Build GRCh38
rs1000687972 Chr.7: 100089004-100089004 on Build GRCh38
rs778348350 Chr.7: 100089045-100089045 on Build GRCh38
rs1045369935 Chr.7: 100088962-100088962 on Build GRCh38
rs900620990 Chr.7: 100089032-100089032 on Build GRCh38
rs1019287496 Chr.7: 100089042-100089042 on Build GRCh38
rs931467459 Chr.7: 100089077-100089077 on Build GRCh38

View Full Product Details