Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165484375-165484644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPE
Entrez Gene ID
1363
Gene Name
carboxypeptidase E
Gene Aliases
CPH
UniGene
Hs.75360
Gene Chromosome Location
Chr.4: 165378942-165498547 on Build GRCh38
Transcripts
NM_001873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3335546 c.797C>T p.A266V Chr.4: 165484428-165484428 on Build GRCh38
5497772 c.945C>T p.N315N Chr.4: 165484576-165484576 on Build GRCh38
3335549 c.802G>A p.E268K Chr.4: 165484433-165484433 on Build GRCh38
4115460 c.905A>C p.K302T Chr.4: 165484536-165484536 on Build GRCh38
6166606 c.949G>T p.G317C Chr.4: 165484580-165484580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4690821 Chr.4: 165484432-165484432 on Build GRCh38
rs553883034 Chr.4: 165484413-165484413 on Build GRCh38
rs987916898 Chr.4: 165484407-165484407 on Build GRCh38
rs142408232 Chr.4: 165484478-165484478 on Build GRCh38
rs184912639 Chr.4: 165484456-165484456 on Build GRCh38
rs771304885 Chr.4: 165484483-165484483 on Build GRCh38
rs202048102 Chr.4: 165484520-165484520 on Build GRCh38
rs775722708 Chr.4: 165484504-165484504 on Build GRCh38
rs148285380 Chr.4: 165484498-165484498 on Build GRCh38
rs746173997 Chr.4: 165484565-165484565 on Build GRCh38
rs753892074 Chr.4: 165484433-165484433 on Build GRCh38
rs775810485 Chr.4: 165484573-165484573 on Build GRCh38
rs773083460 Chr.4: 165484594-165484594 on Build GRCh38
rs764215950 Chr.4: 165484577-165484577 on Build GRCh38
rs145229675 Chr.4: 165484440-165484440 on Build GRCh38
rs150784271 Chr.4: 165484569-165484569 on Build GRCh38
rs141250110 Chr.4: 165484533-165484533 on Build GRCh38
rs543303316 Chr.4: 165484475-165484475 on Build GRCh38
rs201133905 Chr.4: 165484576-165484576 on Build GRCh38
rs372078133 Chr.4: 165484542-165484542 on Build GRCh38
rs760783563 Chr.4: 165484507-165484507 on Build GRCh38
rs774493094 Chr.4: 165484489-165484489 on Build GRCh38
rs138641684 Chr.4: 165484532-165484532 on Build GRCh38
rs767469899 Chr.4: 165484501-165484501 on Build GRCh38
rs763320577 Chr.4: 165484407-165484407 on Build GRCh38
rs202153664 Chr.4: 165484607-165484607 on Build GRCh38
rs771265447 Chr.4: 165484543-165484543 on Build GRCh38
rs774780895 Chr.4: 165484544-165484544 on Build GRCh38
rs137887294 Chr.4: 165484467-165484467 on Build GRCh38
rs1033233885 Chr.4: 165484604-165484604 on Build GRCh38
rs572029817 Chr.4: 165484418-165484418 on Build GRCh38
rs757263530 Chr.4: 165484437-165484437 on Build GRCh38
rs765113959 Chr.4: 165484512-165484512 on Build GRCh38
rs778191319 Chr.4: 165484538-165484538 on Build GRCh38
rs758176349 Chr.4: 165484442-165484442 on Build GRCh38
rs200742682 Chr.4: 165484402-165484402 on Build GRCh38
rs139195612 Chr.4: 165484599-165484599 on Build GRCh38
rs759737700 Chr.4: 165484427-165484427 on Build GRCh38
rs926102003 Chr.4: 165484439-165484439 on Build GRCh38
rs112118467 Chr.4: 165484420-165484420 on Build GRCh38
rs770212500 Chr.4: 165484400-165484400 on Build GRCh38
rs116683740 Chr.4: 165484518-165484518 on Build GRCh38
rs374345764 Chr.4: 165484500-165484500 on Build GRCh38
rs559552538 Chr.4: 165484528-165484528 on Build GRCh38
rs747679979 Chr.4: 165484534-165484534 on Build GRCh38
rs541147146 Chr.4: 165484521-165484521 on Build GRCh38
rs746645558 Chr.4: 165484457-165484457 on Build GRCh38
rs762947355 Chr.4: 165484595-165484595 on Build GRCh38
rs755610315 Chr.4: 165484537-165484537 on Build GRCh38
rs763266873 Chr.4: 165484479-165484479 on Build GRCh38
rs766532383 Chr.4: 165484409-165484409 on Build GRCh38
rs759832878 Chr.4: 165484495-165484495 on Build GRCh38

View Full Product Details