Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36281116-36281349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEPD1
Entrez Gene ID
80820
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
HSPC107
UniGene
Hs.487994
Gene Chromosome Location
Chr.7: 36153149-36301543 on Build GRCh38
Transcripts
NM_030636.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
328208 c.947A>C p.N316T Chr.7: 36281131-36281131 on Build GRCh38
7105742 c.1015G>A p.E339K Chr.7: 36281199-36281199 on Build GRCh38
6258045 c.1009G>T p.V337F Chr.7: 36281193-36281193 on Build GRCh38
7256161 c.963C>A p.P321P Chr.7: 36281147-36281147 on Build GRCh38
3638419 c.989G>C p.R330P Chr.7: 36281173-36281173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564346625 Chr.7: 36281322-36281322 on Build GRCh38
rs112634869 Chr.7: 36281189-36281189 on Build GRCh38
rs761971193 Chr.7: 36281219-36281219 on Build GRCh38
rs779933198 Chr.7: 36281167-36281167 on Build GRCh38
rs1028689831 Chr.7: 36281154-36281154 on Build GRCh38
rs750540337 Chr.7: 36281238-36281238 on Build GRCh38
rs756142246 Chr.7: 36281165-36281165 on Build GRCh38
rs536659570 Chr.7: 36281173-36281173 on Build GRCh38
rs760619832 Chr.7: 36281243-36281243 on Build GRCh38
rs767324515 Chr.7: 36281155-36281155 on Build GRCh38
rs777271692 Chr.7: 36281276-36281276 on Build GRCh38
rs1019797143 Chr.7: 36281199-36281199 on Build GRCh38
rs751579595 Chr.7: 36281148-36281148 on Build GRCh38
rs140756087 Chr.7: 36281147-36281147 on Build GRCh38
rs367555599 Chr.7: 36281275-36281275 on Build GRCh38
rs148018632 Chr.7: 36281138-36281138 on Build GRCh38
rs770290299 Chr.7: 36281209-36281209 on Build GRCh38
rs34655062 Chr.7: 36281194-36281194 on Build GRCh38
rs763926104 Chr.7: 36281274-36281275 on Build GRCh38
rs766488266 Chr.7: 36281249-36281249 on Build GRCh38
rs778606403 Chr.7: 36281271-36281271 on Build GRCh38
rs196586 Chr.7: 36281212-36281212 on Build GRCh38
rs754844279 Chr.7: 36281267-36281267 on Build GRCh38
rs150246825 Chr.7: 36281190-36281190 on Build GRCh38
rs116883157 Chr.7: 36281324-36281324 on Build GRCh38
rs763367291 Chr.7: 36281210-36281210 on Build GRCh38
rs189821881 Chr.7: 36281282-36281282 on Build GRCh38
rs368250302 Chr.7: 36281234-36281234 on Build GRCh38
rs374219442 Chr.7: 36281172-36281172 on Build GRCh38
rs545168097 Chr.7: 36281296-36281296 on Build GRCh38
rs746316571 Chr.7: 36281205-36281205 on Build GRCh38
rs754669971 Chr.7: 36281170-36281170 on Build GRCh38
rs753687138 Chr.7: 36281168-36281168 on Build GRCh38
rs762000997 Chr.7: 36281149-36281149 on Build GRCh38
rs990853779 Chr.7: 36281313-36281313 on Build GRCh38
rs375693705 Chr.7: 36281272-36281272 on Build GRCh38
rs574358346 Chr.7: 36281137-36281137 on Build GRCh38
rs753670691 Chr.7: 36281266-36281266 on Build GRCh38
rs781338969 Chr.7: 36281216-36281216 on Build GRCh38

View Full Product Details