Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31180909-31181158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN14
Entrez Gene ID
440854
Gene Name
calpain 14
Gene Aliases
-
UniGene
Hs.468059
Gene Chromosome Location
Chr.2: 31173056-31233957 on Build GRCh38
Transcripts
NM_001145122.1 NM_001321270.1 XM_011532863.2 XM_011532864.2 XM_011532865.1 XM_011532866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4530354 c.1690G>A p.G564R Chr.2: 31180956-31180956 on Build GRCh38
2941735 c.1710+1G>A p.? Chr.2: 31180935-31180935 on Build GRCh38
4312775 c.1710+1G>A p.? Chr.2: 31180935-31180935 on Build GRCh38
4530353 c.1690G>A p.G564R Chr.2: 31180956-31180956 on Build GRCh38
4312779 c.1655G>A p.S552N Chr.2: 31180991-31180991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950598563 Chr.2: 31181136-31181136 on Build GRCh38
rs764613923 Chr.2: 31180936-31180936 on Build GRCh38
rs878924278 Chr.2: 31181012-31181012 on Build GRCh38
rs1010514728 Chr.2: 31181114-31181127 on Build GRCh38
rs766475493 Chr.2: 31181021-31181021 on Build GRCh38
rs946792299 Chr.2: 31181114-31181114 on Build GRCh38
rs2028677 Chr.2: 31181135-31181135 on Build GRCh38
rs906296159 Chr.2: 31181046-31181046 on Build GRCh38
rs977927780 Chr.2: 31181118-31181118 on Build GRCh38
rs961593805 Chr.2: 31181060-31181060 on Build GRCh38
rs748893120 Chr.2: 31180942-31180942 on Build GRCh38
rs781002406 Chr.2: 31181047-31181047 on Build GRCh38
rs913463697 Chr.2: 31180999-31180999 on Build GRCh38
rs372449814 Chr.2: 31180991-31180991 on Build GRCh38
rs902389007 Chr.2: 31180973-31180973 on Build GRCh38
rs754234522 Chr.2: 31180951-31180951 on Build GRCh38
rs377551095 Chr.2: 31181049-31181049 on Build GRCh38
rs201159308 Chr.2: 31181009-31181009 on Build GRCh38
rs887532555 Chr.2: 31181041-31181041 on Build GRCh38
rs145715824 Chr.2: 31181095-31181095 on Build GRCh38
rs765924190 Chr.2: 31181121-31181134 on Build GRCh38
rs755168692 Chr.2: 31181038-31181038 on Build GRCh38
rs199964386 Chr.2: 31180996-31180996 on Build GRCh38
rs751785811 Chr.2: 31181025-31181025 on Build GRCh38
rs1042620581 Chr.2: 31180958-31180958 on Build GRCh38
rs543956945 Chr.2: 31180982-31180982 on Build GRCh38
rs1035293480 Chr.2: 31181090-31181090 on Build GRCh38
rs915293019 Chr.2: 31181113-31181113 on Build GRCh38
rs73921565 Chr.2: 31181054-31181054 on Build GRCh38
rs1027398700 Chr.2: 31181027-31181027 on Build GRCh38
rs773883138 Chr.2: 31180935-31180935 on Build GRCh38
rs991259862 Chr.2: 31181100-31181100 on Build GRCh38
rs76939962 Chr.2: 31181013-31181013 on Build GRCh38
rs762451735 Chr.2: 31180969-31180969 on Build GRCh38
rs2028678 Chr.2: 31181081-31181081 on Build GRCh38
rs773122364 Chr.2: 31181091-31181091 on Build GRCh38
rs548562879 Chr.2: 31181078-31181078 on Build GRCh38

View Full Product Details