Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176643193-176643458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E1B
Entrez Gene ID
253314
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases
-
UniGene
Hs.448362
Gene Chromosome Location
Chr.5: 176630613-176646644 on Build GRCh38
Transcripts
NM_001099408.1 XM_011534499.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7000163 c.148C>T p.R50W Chr.5: 176643214-176643214 on Build GRCh38
332993 c.148C>A p.R50R Chr.5: 176643214-176643214 on Build GRCh38
3340566 c.153G>A p.T51T Chr.5: 176643219-176643219 on Build GRCh38
6170567 c.162C>A p.P54P Chr.5: 176643228-176643228 on Build GRCh38
391580 c.153delG p.G53fs*110 Chr.5: 176643219-176643219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779126438 Chr.5: 176643218-176643218 on Build GRCh38
rs768843636 Chr.5: 176643298-176643298 on Build GRCh38
rs879893710 Chr.5: 176643325-176643325 on Build GRCh38
rs191363969 Chr.5: 176643375-176643375 on Build GRCh38
rs757549714 Chr.5: 176643275-176643275 on Build GRCh38
rs764428136 Chr.5: 176643271-176643271 on Build GRCh38
rs972111239 Chr.5: 176643411-176643411 on Build GRCh38
rs775973116 Chr.5: 176643221-176643221 on Build GRCh38
rs375992710 Chr.5: 176643225-176643225 on Build GRCh38
rs538159218 Chr.5: 176643397-176643397 on Build GRCh38
rs772701144 Chr.5: 176643229-176643229 on Build GRCh38
rs372956251 Chr.5: 176643219-176643219 on Build GRCh38
rs762984605 Chr.5: 176643230-176643230 on Build GRCh38
rs1010752180 Chr.5: 176643344-176643344 on Build GRCh38
rs920713529 Chr.5: 176643412-176643412 on Build GRCh38
rs751404509 Chr.5: 176643259-176643259 on Build GRCh38
rs370663962 Chr.5: 176643273-176643273 on Build GRCh38
rs376927179 Chr.5: 176643300-176643300 on Build GRCh38
rs769402313 Chr.5: 176643315-176643315 on Build GRCh38
rs117586833 Chr.5: 176643215-176643215 on Build GRCh38
rs747403856 Chr.5: 176643293-176643293 on Build GRCh38
rs972162057 Chr.5: 176643213-176643213 on Build GRCh38
rs758969710 Chr.5: 176643285-176643285 on Build GRCh38
rs991569050 Chr.5: 176643303-176643303 on Build GRCh38
rs368807408 Chr.5: 176643349-176643349 on Build GRCh38
rs942366045 Chr.5: 176643250-176643250 on Build GRCh38
rs747289836 Chr.5: 176643223-176643223 on Build GRCh38
rs1048302519 Chr.5: 176643228-176643228 on Build GRCh38
rs114765382 Chr.5: 176643387-176643387 on Build GRCh38
rs760493029 Chr.5: 176643270-176643282 on Build GRCh38
rs1034996320 Chr.5: 176643395-176643395 on Build GRCh38
rs372424101 Chr.5: 176643357-176643357 on Build GRCh38
rs751393132 Chr.5: 176643218-176643219 on Build GRCh38
rs780180060 Chr.5: 176643224-176643224 on Build GRCh38
rs373176653 Chr.5: 176643283-176643283 on Build GRCh38
rs962054668 Chr.5: 176643398-176643398 on Build GRCh38
rs931150747 Chr.5: 176643425-176643425 on Build GRCh38
rs780383851 Chr.5: 176643286-176643286 on Build GRCh38
rs1040708758 Chr.5: 176643264-176643264 on Build GRCh38
rs749386549 Chr.5: 176643214-176643214 on Build GRCh38
rs79427245 Chr.5: 176643435-176643435 on Build GRCh38
rs779175353 Chr.5: 176643282-176643282 on Build GRCh38
rs766202600 Chr.5: 176643249-176643249 on Build GRCh38
rs747859531 Chr.5: 176643307-176643307 on Build GRCh38
rs754665422 Chr.5: 176643266-176643266 on Build GRCh38

View Full Product Details