Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35319818-35320069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEF6
Entrez Gene ID
50619
Gene Name
DEF6, guanine nucleotide exchange factor
Gene Aliases
IBP, SLAT, SWAP70L
UniGene
Hs.15476
Gene Chromosome Location
Chr.6: 35297818-35321771 on Build GRCh38
Transcripts
NM_022047.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4601312 c.1393G>T p.E465* Chr.6: 35319829-35319829 on Build GRCh38
127791 c.1569C>T p.D523D Chr.6: 35320005-35320005 on Build GRCh38
6642150 c.1445G>A p.R482H Chr.6: 35319881-35319881 on Build GRCh38
5654070 c.1542G>A p.E514E Chr.6: 35319978-35319978 on Build GRCh38
1621563 c.1452C>A p.I484I Chr.6: 35319888-35319888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35964122 Chr.6: 35319890-35319891 on Build GRCh38
rs748091203 Chr.6: 35319991-35319991 on Build GRCh38
rs757628782 Chr.6: 35319880-35319880 on Build GRCh38
rs781478634 Chr.6: 35319886-35319886 on Build GRCh38
rs141385234 Chr.6: 35319885-35319885 on Build GRCh38
rs764630979 Chr.6: 35319860-35319860 on Build GRCh38
rs746250643 Chr.6: 35319888-35319888 on Build GRCh38
rs865793540 Chr.6: 35319893-35319893 on Build GRCh38
rs61734578 Chr.6: 35319978-35319978 on Build GRCh38
rs768317306 Chr.6: 35319983-35319983 on Build GRCh38
rs979363326 Chr.6: 35320007-35320007 on Build GRCh38
rs779878203 Chr.6: 35319944-35319944 on Build GRCh38
rs775847885 Chr.6: 35319961-35319961 on Build GRCh38
rs774321183 Chr.6: 35319984-35319984 on Build GRCh38
rs114708018 Chr.6: 35320034-35320034 on Build GRCh38
rs752715194 Chr.6: 35319849-35319849 on Build GRCh38
rs771895537 Chr.6: 35319998-35319998 on Build GRCh38
rs530306182 Chr.6: 35320025-35320025 on Build GRCh38
rs892058874 Chr.6: 35319896-35319896 on Build GRCh38
rs1011724637 Chr.6: 35319937-35319937 on Build GRCh38
rs547573694 Chr.6: 35319873-35319873 on Build GRCh38
rs775731036 Chr.6: 35320040-35320040 on Build GRCh38
rs758504645 Chr.6: 35319859-35319859 on Build GRCh38
rs755774531 Chr.6: 35319942-35319942 on Build GRCh38

View Full Product Details