Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17027191-17027424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU3
Entrez Gene ID
286097
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
UniGene
Hs.403594
Gene Chromosome Location
Chr.8: 17027238-17133069 on Build GRCh38
Transcripts
NM_181723.2 XM_005273477.2 XM_006716328.2 XM_006716329.2 XM_006716330.2 XM_006716333.3 XM_011544507.2 XM_011544508.2 XM_017013336.1 XM_017013337.1 XM_017013338.1 XM_017013339.1 XM_017013340.1 XM_017013341.1 XM_017013342.1 XM_017013343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7264379 c.8C>T p.A3V Chr.8: 17027287-17027287 on Build GRCh38
3898472 c.89G>A p.R30Q Chr.8: 17027368-17027368 on Build GRCh38
6712664 c.1A>G p.M1V Chr.8: 17027280-17027280 on Build GRCh38
6180453 c.62A>C p.H21P Chr.8: 17027341-17027341 on Build GRCh38
1455810 c.88C>T p.R30W Chr.8: 17027367-17027367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372253445 Chr.8: 17027249-17027249 on Build GRCh38
rs550558198 Chr.8: 17027398-17027398 on Build GRCh38
rs2705203 Chr.8: 17027368-17027368 on Build GRCh38
rs746673506 Chr.8: 17027376-17027376 on Build GRCh38
rs200397681 Chr.8: 17027387-17027387 on Build GRCh38
rs747414231 Chr.8: 17027257-17027257 on Build GRCh38
rs755944140 Chr.8: 17027251-17027251 on Build GRCh38
rs768201460 Chr.8: 17027378-17027378 on Build GRCh38
rs932312943 Chr.8: 17027246-17027246 on Build GRCh38
rs772544891 Chr.8: 17027235-17027235 on Build GRCh38
rs11996774 Chr.8: 17027221-17027221 on Build GRCh38
rs776057777 Chr.8: 17027236-17027236 on Build GRCh38
rs773790027 Chr.8: 17027295-17027295 on Build GRCh38
rs968432025 Chr.8: 17027360-17027360 on Build GRCh38
rs199831377 Chr.8: 17027287-17027287 on Build GRCh38
rs768085292 Chr.8: 17027238-17027239 on Build GRCh38
rs766419108 Chr.8: 17027328-17027328 on Build GRCh38
rs769211097 Chr.8: 17027239-17027239 on Build GRCh38
rs992682130 Chr.8: 17027388-17027388 on Build GRCh38
rs767292347 Chr.8: 17027250-17027250 on Build GRCh38
rs746136994 Chr.8: 17027234-17027234 on Build GRCh38
rs548448436 Chr.8: 17027255-17027255 on Build GRCh38
rs780345689 Chr.8: 17027277-17027277 on Build GRCh38
rs768994977 Chr.8: 17027259-17027259 on Build GRCh38
rs772491468 Chr.8: 17027256-17027256 on Build GRCh38
rs762556432 Chr.8: 17027242-17027242 on Build GRCh38
rs760297492 Chr.8: 17027302-17027302 on Build GRCh38
rs763839697 Chr.8: 17027268-17027268 on Build GRCh38
rs758636723 Chr.8: 17027276-17027276 on Build GRCh38
rs769814462 Chr.8: 17027383-17027383 on Build GRCh38
rs753795183 Chr.8: 17027269-17027269 on Build GRCh38
rs939494293 Chr.8: 17027219-17027220 on Build GRCh38
rs760545798 Chr.8: 17027267-17027267 on Build GRCh38
rs376287896 Chr.8: 17027213-17027218 on Build GRCh38
rs761724049 Chr.8: 17027381-17027381 on Build GRCh38
rs770288058 Chr.8: 17027290-17027290 on Build GRCh38
rs773260307 Chr.8: 17027384-17027384 on Build GRCh38
rs765405139 Chr.8: 17027270-17027270 on Build GRCh38
rs761777555 Chr.8: 17027316-17027316 on Build GRCh38
rs771593237 Chr.8: 17027365-17027365 on Build GRCh38
rs767998769 Chr.8: 17027360-17027361 on Build GRCh38
rs777163774 Chr.8: 17027260-17027260 on Build GRCh38
rs781551166 Chr.8: 17027347-17027347 on Build GRCh38
rs778280523 Chr.8: 17027352-17027352 on Build GRCh38
rs749829944 Chr.8: 17027353-17027353 on Build GRCh38
rs775261435 Chr.8: 17027238-17027245 on Build GRCh38
rs769629251 Chr.8: 17027235-17027245 on Build GRCh38
rs751793250 Chr.8: 17027335-17027335 on Build GRCh38
rs568147819 Chr.8: 17027369-17027369 on Build GRCh38
rs772539961 Chr.8: 17027240-17027240 on Build GRCh38
rs767167386 Chr.8: 17027264-17027264 on Build GRCh38
rs756560810 Chr.8: 17027349-17027349 on Build GRCh38
rs867308812 Chr.8: 17027216-17027216 on Build GRCh38
rs755220439 Chr.8: 17027341-17027341 on Build GRCh38
rs773916489 Chr.8: 17027262-17027262 on Build GRCh38
rs759742770 Chr.8: 17027404-17027404 on Build GRCh38
rs745401676 Chr.8: 17027296-17027296 on Build GRCh38
rs903480573 Chr.8: 17027243-17027243 on Build GRCh38
rs375316594 Chr.8: 17027265-17027265 on Build GRCh38
rs866288006 Chr.8: 17027224-17027224 on Build GRCh38
rs1049475648 Chr.8: 17027247-17027247 on Build GRCh38
rs776637688 Chr.8: 17027314-17027314 on Build GRCh38
rs751593608 Chr.8: 17027401-17027401 on Build GRCh38
rs748873254 Chr.8: 17027238-17027240 on Build GRCh38
rs747235012 Chr.8: 17027285-17027285 on Build GRCh38
rs761083943 Chr.8: 17027238-17027238 on Build GRCh38
rs914080569 Chr.8: 17027229-17027229 on Build GRCh38
rs777784151 Chr.8: 17027253-17027253 on Build GRCh38
rs781466293 Chr.8: 17027286-17027286 on Build GRCh38
rs369005531 Chr.8: 17027272-17027272 on Build GRCh38
rs775181372 Chr.8: 17027297-17027297 on Build GRCh38
rs748523929 Chr.8: 17027348-17027348 on Build GRCh38
rs770412789 Chr.8: 17027261-17027261 on Build GRCh38
rs956751390 Chr.8: 17027275-17027275 on Build GRCh38
rs374933358 Chr.8: 17027342-17027342 on Build GRCh38
rs751189536 Chr.8: 17027245-17027245 on Build GRCh38
rs763938987 Chr.8: 17027311-17027311 on Build GRCh38
rs867640483 Chr.8: 17027218-17027218 on Build GRCh38
rs966272199 Chr.8: 17027385-17027385 on Build GRCh38
rs765913440 Chr.8: 17027244-17027244 on Build GRCh38
rs773865220 Chr.8: 17027242-17027245 on Build GRCh38
rs986186451 Chr.8: 17027362-17027362 on Build GRCh38
rs766474206 Chr.8: 17027399-17027399 on Build GRCh38
rs762242856 Chr.8: 17027244-17027245 on Build GRCh38
rs771223954 Chr.8: 17027230-17027230 on Build GRCh38
rs750483107 Chr.8: 17027317-17027317 on Build GRCh38
rs757385684 Chr.8: 17027254-17027254 on Build GRCh38

View Full Product Details