Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6477066-6477331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP2
Entrez Gene ID
23647
Gene Name
ADP ribosylation factor interacting protein 2
Gene Aliases
POR1
UniGene
Hs.732203
Gene Chromosome Location
Chr.11: 6474683-6481479 on Build GRCh38
Transcripts
NM_001242854.1 NM_001242855.1 NM_001242856.1 NM_012402.3 XM_005252840.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4838226 c.982C>T p.R328W Chr.11: 6477157-6477157 on Build GRCh38
5970285 c.958C>G p.L320V Chr.11: 6477181-6477181 on Build GRCh38
234406 c.936C>T p.N312N Chr.11: 6477203-6477203 on Build GRCh38
5670999 c.881T>C p.M294T Chr.11: 6477258-6477258 on Build GRCh38
5798614 c.987T>G p.P329P Chr.11: 6477152-6477152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866955358 Chr.11: 6477190-6477190 on Build GRCh38
rs751330094 Chr.11: 6477152-6477152 on Build GRCh38
rs767165568 Chr.11: 6477299-6477299 on Build GRCh38
rs532928941 Chr.11: 6477221-6477221 on Build GRCh38
rs372336395 Chr.11: 6477222-6477222 on Build GRCh38
rs375721029 Chr.11: 6477241-6477241 on Build GRCh38
rs762696754 Chr.11: 6477257-6477257 on Build GRCh38
rs772772225 Chr.11: 6477245-6477245 on Build GRCh38
rs368145555 Chr.11: 6477156-6477156 on Build GRCh38
rs759846347 Chr.11: 6477218-6477218 on Build GRCh38
rs765682112 Chr.11: 6477235-6477235 on Build GRCh38
rs370684963 Chr.11: 6477167-6477167 on Build GRCh38
rs760042625 Chr.11: 6477091-6477091 on Build GRCh38
rs758109695 Chr.11: 6477149-6477149 on Build GRCh38
rs150607280 Chr.11: 6477112-6477112 on Build GRCh38
rs374345673 Chr.11: 6477181-6477181 on Build GRCh38
rs751418132 Chr.11: 6477286-6477286 on Build GRCh38
rs764772903 Chr.11: 6477132-6477132 on Build GRCh38
rs201024768 Chr.11: 6477308-6477308 on Build GRCh38
rs761222248 Chr.11: 6477239-6477239 on Build GRCh38
rs141798060 Chr.11: 6477234-6477234 on Build GRCh38
rs761188444 Chr.11: 6477131-6477131 on Build GRCh38
rs773493084 Chr.11: 6477157-6477157 on Build GRCh38
rs780938078 Chr.11: 6477160-6477160 on Build GRCh38
rs368728080 Chr.11: 6477211-6477211 on Build GRCh38
rs748286505 Chr.11: 6477166-6477166 on Build GRCh38
rs771077067 Chr.11: 6477195-6477195 on Build GRCh38
rs763234616 Chr.11: 6477277-6477277 on Build GRCh38
rs571039502 Chr.11: 6477306-6477306 on Build GRCh38
rs775860173 Chr.11: 6477242-6477242 on Build GRCh38
rs767955114 Chr.11: 6477093-6477093 on Build GRCh38
rs780059833 Chr.11: 6477150-6477150 on Build GRCh38
rs749581427 Chr.11: 6477188-6477188 on Build GRCh38
rs750091716 Chr.11: 6477134-6477134 on Build GRCh38

View Full Product Details