Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75128640-75128887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC7
Entrez Gene ID
79637
Gene Name
armadillo repeat containing 7
Gene Aliases
-
UniGene
Hs.413047
Gene Chromosome Location
Chr.17: 75109952-75130265 on Build GRCh38
Transcripts
NM_001304271.1 NM_024585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069491 c.390C>G p.R130R Chr.17: 75128831-75128831 on Build GRCh38
328419 c.384G>A p.P128P Chr.17: 75128825-75128825 on Build GRCh38
3820527 c.256C>A p.P86T Chr.17: 75128697-75128697 on Build GRCh38
327373 c.259G>A p.D87N Chr.17: 75128700-75128700 on Build GRCh38
4663410 c.252G>T p.L84L Chr.17: 75128693-75128693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377200687 Chr.17: 75128822-75128822 on Build GRCh38
rs575470135 Chr.17: 75128715-75128715 on Build GRCh38
rs371615008 Chr.17: 75128829-75128829 on Build GRCh38
rs1008439883 Chr.17: 75128690-75128690 on Build GRCh38
rs760643043 Chr.17: 75128682-75128682 on Build GRCh38
rs777603204 Chr.17: 75128764-75128764 on Build GRCh38
rs536253540 Chr.17: 75128703-75128703 on Build GRCh38
rs774740585 Chr.17: 75128816-75128816 on Build GRCh38
rs774711121 Chr.17: 75128861-75128861 on Build GRCh38
rs766956073 Chr.17: 75128812-75128812 on Build GRCh38
rs367787212 Chr.17: 75128840-75128840 on Build GRCh38
rs763624572 Chr.17: 75128826-75128826 on Build GRCh38
rs943437324 Chr.17: 75128732-75128734 on Build GRCh38
rs936024956 Chr.17: 75128670-75128670 on Build GRCh38
rs978422777 Chr.17: 75128844-75128844 on Build GRCh38
rs755552156 Chr.17: 75128867-75128867 on Build GRCh38
rs750539776 Chr.17: 75128758-75128766 on Build GRCh38
rs995452833 Chr.17: 75128762-75128762 on Build GRCh38
rs188117376 Chr.17: 75128730-75128730 on Build GRCh38
rs770178998 Chr.17: 75128856-75128856 on Build GRCh38
rs768891176 Chr.17: 75128789-75128789 on Build GRCh38
rs780632024 Chr.17: 75128739-75128739 on Build GRCh38
rs761904572 Chr.17: 75128839-75128839 on Build GRCh38
rs199560912 Chr.17: 75128851-75128851 on Build GRCh38
rs138342781 Chr.17: 75128729-75128729 on Build GRCh38
rs755705792 Chr.17: 75128763-75128763 on Build GRCh38
rs768008219 Chr.17: 75128862-75128862 on Build GRCh38
rs993670784 Chr.17: 75128761-75128761 on Build GRCh38
rs778733298 Chr.17: 75128833-75128833 on Build GRCh38
rs377525655 Chr.17: 75128821-75128821 on Build GRCh38
rs778696108 Chr.17: 75128782-75128782 on Build GRCh38
rs113871549 Chr.17: 75128830-75128830 on Build GRCh38
rs888026059 Chr.17: 75128780-75128780 on Build GRCh38
rs866708507 Chr.17: 75128807-75128807 on Build GRCh38
rs150451517 Chr.17: 75128704-75128704 on Build GRCh38
rs184299476 Chr.17: 75128678-75128678 on Build GRCh38
rs754422264 Chr.17: 75128734-75128734 on Build GRCh38
rs769050629 Chr.17: 75128683-75128683 on Build GRCh38
rs748230926 Chr.17: 75128854-75128854 on Build GRCh38
rs770943213 Chr.17: 75128778-75128778 on Build GRCh38
rs201640079 Chr.17: 75128773-75128773 on Build GRCh38
rs374083928 Chr.17: 75128746-75128748 on Build GRCh38
rs150791316 Chr.17: 75128798-75128798 on Build GRCh38
rs1035491706 Chr.17: 75128811-75128811 on Build GRCh38
rs750693086 Chr.17: 75128720-75128720 on Build GRCh38
rs145777741 Chr.17: 75128808-75128808 on Build GRCh38
rs752377953 Chr.17: 75128753-75128753 on Build GRCh38
rs766789681 Chr.17: 75128727-75128727 on Build GRCh38
rs199960480 Chr.17: 75128853-75128853 on Build GRCh38
rs771230796 Chr.17: 75128858-75128858 on Build GRCh38
rs529431052 Chr.17: 75128828-75128828 on Build GRCh38
rs752190084 Chr.17: 75128824-75128824 on Build GRCh38
rs1040436516 Chr.17: 75128735-75128735 on Build GRCh38
rs762220396 Chr.17: 75128809-75128809 on Build GRCh38
rs200997688 Chr.17: 75128825-75128825 on Build GRCh38
rs142697875 Chr.17: 75128857-75128857 on Build GRCh38
rs770124022 Chr.17: 75128803-75128803 on Build GRCh38
rs745772379 Chr.17: 75128784-75128784 on Build GRCh38
rs773689492 Chr.17: 75128804-75128804 on Build GRCh38
rs368397295 Chr.17: 75128723-75128723 on Build GRCh38
rs758349108 Chr.17: 75128852-75128852 on Build GRCh38

View Full Product Details