Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33174030-33174265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA2
Entrez Gene ID
140683
Gene Name
BPI fold containing family A member 2
Gene Aliases
C20orf70, PSP, SPLUNC2, bA49G10.1
UniGene
Hs.125960
Gene Chromosome Location
Chr.20: 33161848-33181417 on Build GRCh38
Transcripts
NM_001319164.1 NM_080574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3991642 c.341C>T p.A114V Chr.20: 33174117-33174117 on Build GRCh38
3363191 c.315C>A p.S105R Chr.20: 33174091-33174091 on Build GRCh38
3911120 c.371A>C p.N124T Chr.20: 33174147-33174147 on Build GRCh38
4665961 c.352G>A p.D118N Chr.20: 33174128-33174128 on Build GRCh38
5602920 c.373C>G p.L125V Chr.20: 33174149-33174149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565760495 Chr.20: 33174101-33174101 on Build GRCh38
rs767366080 Chr.20: 33174067-33174067 on Build GRCh38
rs764574585 Chr.20: 33174178-33174178 on Build GRCh38
rs61734343 Chr.20: 33174079-33174079 on Build GRCh38
rs779811024 Chr.20: 33174072-33174072 on Build GRCh38
rs755982612 Chr.20: 33174166-33174166 on Build GRCh38
rs750167611 Chr.20: 33174162-33174162 on Build GRCh38
rs773237168 Chr.20: 33174226-33174226 on Build GRCh38
rs79337613 Chr.20: 33174197-33174197 on Build GRCh38
rs367714865 Chr.20: 33174074-33174074 on Build GRCh38
rs760561107 Chr.20: 33174227-33174227 on Build GRCh38
rs554973504 Chr.20: 33174169-33174169 on Build GRCh38
rs762039054 Chr.20: 33174216-33174216 on Build GRCh38
rs775821860 Chr.20: 33174116-33174116 on Build GRCh38
rs757749859 Chr.20: 33174185-33174185 on Build GRCh38
rs981420494 Chr.20: 33174123-33174123 on Build GRCh38
rs200274615 Chr.20: 33174069-33174069 on Build GRCh38
rs17304572 Chr.20: 33174113-33174113 on Build GRCh38
rs781748111 Chr.20: 33174187-33174187 on Build GRCh38
rs755714792 Chr.20: 33174071-33174071 on Build GRCh38
rs974095211 Chr.20: 33174140-33174140 on Build GRCh38
rs751505897 Chr.20: 33174056-33174056 on Build GRCh38
rs577845506 Chr.20: 33174239-33174239 on Build GRCh38
rs763143194 Chr.20: 33174055-33174055 on Build GRCh38
rs151315203 Chr.20: 33174184-33174184 on Build GRCh38
rs373907221 Chr.20: 33174168-33174168 on Build GRCh38
rs766147977 Chr.20: 33174235-33174235 on Build GRCh38
rs79185071 Chr.20: 33174237-33174237 on Build GRCh38
rs774565359 Chr.20: 33174214-33174214 on Build GRCh38
rs776857886 Chr.20: 33174108-33174108 on Build GRCh38
rs142458594 Chr.20: 33174149-33174149 on Build GRCh38
rs774589612 Chr.20: 33174134-33174134 on Build GRCh38
rs1021929613 Chr.20: 33174244-33174244 on Build GRCh38
rs376984515 Chr.20: 33174236-33174236 on Build GRCh38
rs770156161 Chr.20: 33174195-33174195 on Build GRCh38
rs764814958 Chr.20: 33174065-33174065 on Build GRCh38
rs769007336 Chr.20: 33174199-33174199 on Build GRCh38
rs145588274 Chr.20: 33174087-33174087 on Build GRCh38
rs367896836 Chr.20: 33174189-33174189 on Build GRCh38
rs771439064 Chr.20: 33174091-33174091 on Build GRCh38
rs149183452 Chr.20: 33174127-33174127 on Build GRCh38
rs771878720 Chr.20: 33174221-33174221 on Build GRCh38
rs764223690 Chr.20: 33174128-33174128 on Build GRCh38
rs987090897 Chr.20: 33174105-33174105 on Build GRCh38

View Full Product Details