Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19973176-19973433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
UniGene
Hs.713616
Gene Chromosome Location
Chr.22: 19966491-20016802 on Build GRCh38
Transcripts
NM_001670.2 XM_005261242.2 XM_005261243.3 XM_005261244.3 XM_006724243.2 XM_006724245.2 XM_006724246.3 XM_006724247.3 XM_006724248.3 XM_011530179.2 XM_011530180.1 XM_011530182.2 XM_011530183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3708141 c.2292G>A p.P764P Chr.22: 19973265-19973265 on Build GRCh38
1740771 c.2357T>C p.I786T Chr.22: 19973200-19973200 on Build GRCh38
5858921 c.2353G>A p.E785K Chr.22: 19973204-19973204 on Build GRCh38
1032163 c.2370C>T p.S790S Chr.22: 19973187-19973187 on Build GRCh38
1283522 c.2379C>T p.N793N Chr.22: 19973178-19973178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781574583 Chr.22: 19973274-19973275 on Build GRCh38
rs540490780 Chr.22: 19973387-19973387 on Build GRCh38
rs780649546 Chr.22: 19973324-19973324 on Build GRCh38
rs757745205 Chr.22: 19973264-19973265 on Build GRCh38
rs779602144 Chr.22: 19973334-19973334 on Build GRCh38
rs1052097389 Chr.22: 19973392-19973392 on Build GRCh38
rs757531262 Chr.22: 19973220-19973220 on Build GRCh38
rs773519155 Chr.22: 19973349-19973349 on Build GRCh38
rs747443430 Chr.22: 19973250-19973250 on Build GRCh38
rs574936740 Chr.22: 19973372-19973372 on Build GRCh38
rs531297219 Chr.22: 19973405-19973405 on Build GRCh38
rs757094742 Chr.22: 19973323-19973323 on Build GRCh38
rs772492250 Chr.22: 19973345-19973345 on Build GRCh38
rs753880291 Chr.22: 19973362-19973362 on Build GRCh38
rs766479611 Chr.22: 19973274-19973274 on Build GRCh38
rs779630375 Chr.22: 19973412-19973412 on Build GRCh38
rs1015405676 Chr.22: 19973390-19973390 on Build GRCh38
rs777535564 Chr.22: 19973245-19973245 on Build GRCh38
rs760941516 Chr.22: 19973353-19973353 on Build GRCh38
rs745313568 Chr.22: 19973252-19973252 on Build GRCh38
rs764556878 Chr.22: 19973216-19973216 on Build GRCh38
rs571153630 Chr.22: 19973397-19973397 on Build GRCh38
rs759123244 Chr.22: 19973200-19973200 on Build GRCh38
rs746878504 Chr.22: 19973236-19973236 on Build GRCh38
rs752037224 Chr.22: 19973217-19973217 on Build GRCh38
rs758305598 Chr.22: 19973288-19973288 on Build GRCh38
rs576793681 Chr.22: 19973235-19973235 on Build GRCh38
rs762300798 Chr.22: 19973265-19973265 on Build GRCh38
rs959806304 Chr.22: 19973264-19973264 on Build GRCh38
rs371730200 Chr.22: 19973263-19973263 on Build GRCh38
rs556648110 Chr.22: 19973251-19973251 on Build GRCh38
rs775022836 Chr.22: 19973337-19973337 on Build GRCh38
rs551628184 Chr.22: 19973395-19973395 on Build GRCh38
rs769399287 Chr.22: 19973326-19973326 on Build GRCh38
rs375012817 Chr.22: 19973382-19973382 on Build GRCh38
rs750674229 Chr.22: 19973269-19973269 on Build GRCh38
rs745537368 Chr.22: 19973325-19973325 on Build GRCh38
rs756242656 Chr.22: 19973233-19973233 on Build GRCh38
rs140904483 Chr.22: 19973258-19973258 on Build GRCh38
rs374908033 Chr.22: 19973256-19973256 on Build GRCh38
rs560919773 Chr.22: 19973335-19973335 on Build GRCh38
rs778661191 Chr.22: 19973234-19973234 on Build GRCh38
rs767822334 Chr.22: 19973227-19973227 on Build GRCh38
rs763549063 Chr.22: 19973260-19973260 on Build GRCh38
rs759639955 Chr.22: 19973366-19973366 on Build GRCh38
rs776085428 Chr.22: 19973205-19973205 on Build GRCh38
rs545917955 Chr.22: 19973248-19973248 on Build GRCh38
rs754803585 Chr.22: 19973328-19973328 on Build GRCh38
rs767587957 Chr.22: 19973266-19973266 on Build GRCh38
rs751312174 Chr.22: 19973306-19973306 on Build GRCh38
rs144187900 Chr.22: 19973204-19973204 on Build GRCh38
rs780920381 Chr.22: 19973344-19973344 on Build GRCh38
rs376268496 Chr.22: 19973272-19973272 on Build GRCh38
rs116570619 Chr.22: 19973199-19973199 on Build GRCh38
rs750431334 Chr.22: 19973231-19973231 on Build GRCh38
rs530016774 Chr.22: 19973368-19973368 on Build GRCh38
rs766563044 Chr.22: 19973360-19973360 on Build GRCh38
rs369590446 Chr.22: 19973278-19973278 on Build GRCh38
rs560203785 Chr.22: 19973393-19973393 on Build GRCh38
rs770623530 Chr.22: 19973198-19973198 on Build GRCh38
rs372539946 Chr.22: 19973291-19973291 on Build GRCh38
rs190544609 Chr.22: 19973404-19973404 on Build GRCh38
rs367914866 Chr.22: 19973262-19973262 on Build GRCh38
rs1043878575 Chr.22: 19973267-19973267 on Build GRCh38
rs146518336 Chr.22: 19973211-19973211 on Build GRCh38

View Full Product Details