Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 108234977-108235206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3E
Entrez Gene ID
3646
Gene Name
eukaryotic translation initiation factor 3 subunit E
Gene Aliases
EIF3-P48, EIF3S6, INT6, eIF3-p46
UniGene
Hs.405590
Gene Chromosome Location
Chr.8: 108201743-108248802 on Build GRCh38
Transcripts
NM_001568.2 XM_017013398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6292161 c.367-8C>T p.? Chr.8: 108235110-108235110 on Build GRCh38
5567426 c.435A>G p.S145S Chr.8: 108235034-108235034 on Build GRCh38
1454032 c.442G>T p.A148S Chr.8: 108235027-108235027 on Build GRCh38
6652834 c.417C>T p.Y139Y Chr.8: 108235052-108235052 on Build GRCh38
1454031 c.462_463insT p.R155fs*1 Chr.8: 108235006-108235007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773810094 Chr.8: 108235158-108235158 on Build GRCh38
rs138258298 Chr.8: 108235052-108235052 on Build GRCh38
rs991719803 Chr.8: 108235111-108235114 on Build GRCh38
rs746371820 Chr.8: 108235029-108235029 on Build GRCh38
rs557435957 Chr.8: 108235155-108235155 on Build GRCh38
rs188624639 Chr.8: 108235096-108235096 on Build GRCh38
rs778335805 Chr.8: 108235059-108235059 on Build GRCh38
rs756546086 Chr.8: 108235153-108235153 on Build GRCh38
rs757720051 Chr.8: 108235057-108235057 on Build GRCh38
rs376289399 Chr.8: 108235064-108235064 on Build GRCh38
rs574034351 Chr.8: 108235167-108235167 on Build GRCh38
rs749060424 Chr.8: 108235148-108235163 on Build GRCh38
rs543063971 Chr.8: 108235171-108235171 on Build GRCh38
rs762968688 Chr.8: 108235115-108235115 on Build GRCh38
rs927297897 Chr.8: 108235070-108235070 on Build GRCh38
rs917522088 Chr.8: 108235135-108235135 on Build GRCh38
rs751701808 Chr.8: 108235131-108235131 on Build GRCh38
rs1099432 Chr.8: 108235110-108235110 on Build GRCh38
rs373892757 Chr.8: 108235076-108235076 on Build GRCh38
rs776633451 Chr.8: 108235095-108235095 on Build GRCh38
rs772758556 Chr.8: 108235074-108235074 on Build GRCh38
rs770815070 Chr.8: 108235091-108235091 on Build GRCh38
rs750798410 Chr.8: 108235148-108235148 on Build GRCh38
rs930620195 Chr.8: 108235138-108235138 on Build GRCh38
rs747609832 Chr.8: 108235034-108235037 on Build GRCh38
rs149619443 Chr.8: 108235177-108235177 on Build GRCh38
rs901645744 Chr.8: 108235175-108235175 on Build GRCh38
rs867145721 Chr.8: 108235066-108235066 on Build GRCh38
rs1104491 Chr.8: 108235109-108235109 on Build GRCh38
rs764283496 Chr.8: 108235121-108235121 on Build GRCh38
rs759491312 Chr.8: 108235104-108235104 on Build GRCh38
rs745400190 Chr.8: 108235111-108235111 on Build GRCh38
rs756024976 Chr.8: 108235133-108235137 on Build GRCh38
rs781578210 Chr.8: 108235028-108235028 on Build GRCh38
rs775603062 Chr.8: 108235113-108235113 on Build GRCh38
rs1040604525 Chr.8: 108235166-108235166 on Build GRCh38
rs780687153 Chr.8: 108235110-108235111 on Build GRCh38
rs771715727 Chr.8: 108235067-108235067 on Build GRCh38
rs767856796 Chr.8: 108235142-108235142 on Build GRCh38
rs775331949 Chr.8: 108235141-108235142 on Build GRCh38
rs769861535 Chr.8: 108235111-108235111 on Build GRCh38
rs762091152 Chr.8: 108235141-108235141 on Build GRCh38
rs746720737 Chr.8: 108235075-108235075 on Build GRCh38

View Full Product Details