Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235454161-235454386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALNT2
Entrez Gene ID
148789
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
B3GalNAc-T2, MDDGA11
UniGene
Hs.498143
Gene Chromosome Location
Chr.1: 235440654-235504481 on Build GRCh38
Transcripts
NM_152490.4 XM_006711749.3 XM_017000394.1 XM_017000395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5506775 c.1174G>C p.D392H Chr.1: 235454293-235454293 on Build GRCh38
4522410 c.1161G>A p.L387L Chr.1: 235454306-235454306 on Build GRCh38
5993467 c.1157G>A p.R386K Chr.1: 235454310-235454310 on Build GRCh38
3485540 c.1260G>A p.K420K Chr.1: 235454207-235454207 on Build GRCh38
6609213 c.1209G>A p.P403P Chr.1: 235454258-235454258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761630404 Chr.1: 235454183-235454183 on Build GRCh38
rs762656574 Chr.1: 235454201-235454201 on Build GRCh38
rs140708018 Chr.1: 235454289-235454289 on Build GRCh38
rs769540544 Chr.1: 235454185-235454185 on Build GRCh38
rs773466522 Chr.1: 235454188-235454188 on Build GRCh38
rs373186743 Chr.1: 235454285-235454285 on Build GRCh38
rs771003959 Chr.1: 235454296-235454296 on Build GRCh38
rs762832960 Chr.1: 235454293-235454293 on Build GRCh38
rs761126849 Chr.1: 235454324-235454324 on Build GRCh38
rs375815861 Chr.1: 235454230-235454230 on Build GRCh38
rs754101273 Chr.1: 235454335-235454335 on Build GRCh38
rs369079608 Chr.1: 235454251-235454251 on Build GRCh38
rs201407732 Chr.1: 235454271-235454271 on Build GRCh38
rs781267171 Chr.1: 235454269-235454269 on Build GRCh38
rs554797006 Chr.1: 235454253-235454253 on Build GRCh38
rs753271269 Chr.1: 235454250-235454250 on Build GRCh38
rs375288669 Chr.1: 235454258-235454258 on Build GRCh38
rs898996974 Chr.1: 235454280-235454280 on Build GRCh38
rs759609345 Chr.1: 235454304-235454304 on Build GRCh38
rs768282637 Chr.1: 235454266-235454266 on Build GRCh38
rs754518690 Chr.1: 235454359-235454359 on Build GRCh38
rs757519127 Chr.1: 235454280-235454281 on Build GRCh38
rs376365276 Chr.1: 235454332-235454332 on Build GRCh38
rs779678584 Chr.1: 235454333-235454335 on Build GRCh38
rs759627815 Chr.1: 235454217-235454217 on Build GRCh38
rs753181340 Chr.1: 235454323-235454323 on Build GRCh38
rs929108412 Chr.1: 235454284-235454284 on Build GRCh38
rs369378819 Chr.1: 235454259-235454259 on Build GRCh38
rs766659361 Chr.1: 235454203-235454203 on Build GRCh38
rs145518904 Chr.1: 235454252-235454252 on Build GRCh38
rs751320018 Chr.1: 235454355-235454355 on Build GRCh38
rs369795107 Chr.1: 235454344-235454344 on Build GRCh38
rs779504258 Chr.1: 235454348-235454348 on Build GRCh38
rs764693057 Chr.1: 235454330-235454330 on Build GRCh38
rs757947010 Chr.1: 235454256-235454256 on Build GRCh38
rs751592765 Chr.1: 235454205-235454205 on Build GRCh38
rs780894404 Chr.1: 235454361-235454361 on Build GRCh38
rs767643583 Chr.1: 235454320-235454320 on Build GRCh38
rs763128655 Chr.1: 235454195-235454195 on Build GRCh38
rs774563506 Chr.1: 235454300-235454300 on Build GRCh38

View Full Product Details