Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78917977-78918232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL4
Entrez Gene ID
64123
Gene Name
adhesion G protein-coupled receptor L4
Gene Aliases
ELTD1, ETL, KPG_003
UniGene
Hs.132314
Gene Chromosome Location
Chr.1: 78889764-79006810 on Build GRCh38
Transcripts
NM_022159.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1344455 c.1513T>C p.F505L Chr.1: 78917999-78917999 on Build GRCh38
3966753 c.1508C>G p.A503G Chr.1: 78918004-78918004 on Build GRCh38
912421 c.1511C>A p.A504D Chr.1: 78918001-78918001 on Build GRCh38
6653579 c.1530T>A p.I510I Chr.1: 78917982-78917982 on Build GRCh38
681989 c.1535G>C p.G512A Chr.1: 78917977-78917977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200740355 Chr.1: 78918064-78918064 on Build GRCh38
rs904263625 Chr.1: 78918109-78918109 on Build GRCh38
rs1008786266 Chr.1: 78918173-78918173 on Build GRCh38
rs757851460 Chr.1: 78918102-78918102 on Build GRCh38
rs773048656 Chr.1: 78918058-78918058 on Build GRCh38
rs759891297 Chr.1: 78918029-78918029 on Build GRCh38
rs891389656 Chr.1: 78918005-78918005 on Build GRCh38
rs75253446 Chr.1: 78918064-78918064 on Build GRCh38
rs764197547 Chr.1: 78918045-78918045 on Build GRCh38
rs753750454 Chr.1: 78918069-78918069 on Build GRCh38
rs146732519 Chr.1: 78918072-78918072 on Build GRCh38
rs575102952 Chr.1: 78918106-78918106 on Build GRCh38
rs768270261 Chr.1: 78918064-78918065 on Build GRCh38
rs140561874 Chr.1: 78918125-78918125 on Build GRCh38
rs747482293 Chr.1: 78918087-78918087 on Build GRCh38
rs766090048 Chr.1: 78918060-78918060 on Build GRCh38
rs917817144 Chr.1: 78918163-78918163 on Build GRCh38
rs775667826 Chr.1: 78918036-78918036 on Build GRCh38
rs200345373 Chr.1: 78918080-78918080 on Build GRCh38
rs777288036 Chr.1: 78918083-78918083 on Build GRCh38
rs770231774 Chr.1: 78918010-78918012 on Build GRCh38
rs78237403 Chr.1: 78918063-78918063 on Build GRCh38
rs771220617 Chr.1: 78918021-78918021 on Build GRCh38
rs763065423 Chr.1: 78918037-78918037 on Build GRCh38
rs908076426 Chr.1: 78918009-78918009 on Build GRCh38
rs572774337 Chr.1: 78918126-78918126 on Build GRCh38
rs867979983 Chr.1: 78918026-78918026 on Build GRCh38
rs17852292 Chr.1: 78918030-78918030 on Build GRCh38
rs531517486 Chr.1: 78918164-78918164 on Build GRCh38
rs760605262 Chr.1: 78918059-78918059 on Build GRCh38
rs778025819 Chr.1: 78918059-78918060 on Build GRCh38
rs72675852 Chr.1: 78918081-78918081 on Build GRCh38
rs765026903 Chr.1: 78918074-78918074 on Build GRCh38
rs774935244 Chr.1: 78918063-78918064 on Build GRCh38
rs776989789 Chr.1: 78918023-78918023 on Build GRCh38
rs979958166 Chr.1: 78918104-78918104 on Build GRCh38
rs139272266 Chr.1: 78918121-78918121 on Build GRCh38

View Full Product Details