Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30187417-30187650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1A
Entrez Gene ID
11151
Gene Name
coronin 1A
Gene Aliases
CLABP, CLIPINA, HCORO1, IMD8, TACO, p57
UniGene
Hs.415067
Gene Chromosome Location
Chr.16: 30183410-30189076 on Build GRCh38
Transcripts
NM_001193333.2 NM_007074.3 XM_011545714.2 XM_017022885.1 XM_017022886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6332923 c.695G>T p.G232V Chr.16: 30187440-30187440 on Build GRCh38
1678997 c.703C>A p.L235M Chr.16: 30187448-30187448 on Build GRCh38
5971755 c.684C>T p.F228F Chr.16: 30187429-30187429 on Build GRCh38
1493636 c.728G>A p.S243N Chr.16: 30187473-30187473 on Build GRCh38
4060094 c.679G>A p.V227M Chr.16: 30187424-30187424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772251544 Chr.16: 30187492-30187492 on Build GRCh38
rs748364715 Chr.16: 30187600-30187600 on Build GRCh38
rs758766497 Chr.16: 30187575-30187575 on Build GRCh38
rs143729497 Chr.16: 30187483-30187483 on Build GRCh38
rs908355576 Chr.16: 30187551-30187551 on Build GRCh38
rs371473241 Chr.16: 30187540-30187540 on Build GRCh38
rs779133797 Chr.16: 30187572-30187572 on Build GRCh38
rs754702749 Chr.16: 30187553-30187553 on Build GRCh38
rs893581339 Chr.16: 30187624-30187624 on Build GRCh38
rs200060318 Chr.16: 30187489-30187489 on Build GRCh38
rs769683287 Chr.16: 30187616-30187616 on Build GRCh38
rs772821967 Chr.16: 30187617-30187617 on Build GRCh38
rs754616768 Chr.16: 30187434-30187434 on Build GRCh38
rs556055190 Chr.16: 30187524-30187524 on Build GRCh38
rs746400107 Chr.16: 30187573-30187573 on Build GRCh38
rs866153700 Chr.16: 30187545-30187545 on Build GRCh38
rs374088140 Chr.16: 30187456-30187456 on Build GRCh38
rs1034209901 Chr.16: 30187470-30187470 on Build GRCh38
rs776285526 Chr.16: 30187511-30187511 on Build GRCh38
rs541262345 Chr.16: 30187595-30187595 on Build GRCh38
rs761645032 Chr.16: 30187523-30187523 on Build GRCh38
rs776430488 Chr.16: 30187513-30187513 on Build GRCh38
rs757710480 Chr.16: 30187564-30187564 on Build GRCh38
rs760892904 Chr.16: 30187512-30187512 on Build GRCh38
rs951171015 Chr.16: 30187518-30187518 on Build GRCh38
rs751043826 Chr.16: 30187544-30187544 on Build GRCh38
rs117041067 Chr.16: 30187529-30187529 on Build GRCh38
rs779518208 Chr.16: 30187485-30187485 on Build GRCh38
rs749622934 Chr.16: 30187465-30187465 on Build GRCh38
rs780215021 Chr.16: 30187576-30187576 on Build GRCh38
rs781043486 Chr.16: 30187560-30187560 on Build GRCh38
rs775376547 Chr.16: 30187507-30187507 on Build GRCh38
rs752370559 Chr.16: 30187561-30187561 on Build GRCh38

View Full Product Details