Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119304254-119304507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf3
Entrez Gene ID
401152
Gene Name
chromosome 4 open reading frame 3
Gene Aliases
-
UniGene
Hs.173705
Gene Chromosome Location
Chr.4: 119296419-119304445 on Build GRCh38
Transcripts
NM_001170330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618328 c.31A>T p.R11W Chr.4: 119304415-119304415 on Build GRCh38
6229509 c.75G>T p.R25S Chr.4: 119304371-119304371 on Build GRCh38
1050467 c.29A>G p.N10S Chr.4: 119304417-119304417 on Build GRCh38
1050466 c.88T>C p.Y30H Chr.4: 119304358-119304358 on Build GRCh38
5622478 c.113G>T p.C38F Chr.4: 119304333-119304333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762803072 Chr.4: 119304430-119304430 on Build GRCh38
rs977778615 Chr.4: 119304472-119304472 on Build GRCh38
rs932033992 Chr.4: 119304377-119304377 on Build GRCh38
rs967388446 Chr.4: 119304325-119304325 on Build GRCh38
rs543679260 Chr.4: 119304354-119304354 on Build GRCh38
rs553091483 Chr.4: 119304356-119304356 on Build GRCh38
rs766444048 Chr.4: 119304441-119304441 on Build GRCh38
rs909336038 Chr.4: 119304394-119304394 on Build GRCh38
rs1025680513 Chr.4: 119304312-119304312 on Build GRCh38
rs913150247 Chr.4: 119304320-119304320 on Build GRCh38
rs552248556 Chr.4: 119304446-119304446 on Build GRCh38
rs769773142 Chr.4: 119304357-119304357 on Build GRCh38
rs772979807 Chr.4: 119304358-119304358 on Build GRCh38
rs942298770 Chr.4: 119304398-119304398 on Build GRCh38
rs990760083 Chr.4: 119304383-119304383 on Build GRCh38
rs115125596 Chr.4: 119304335-119304335 on Build GRCh38
rs561103632 Chr.4: 119304288-119304288 on Build GRCh38
rs768238078 Chr.4: 119304333-119304333 on Build GRCh38
rs749125678 Chr.4: 119304431-119304431 on Build GRCh38
rs750299780 Chr.4: 119304285-119304285 on Build GRCh38
rs1024931322 Chr.4: 119304392-119304392 on Build GRCh38
rs765390076 Chr.4: 119304367-119304367 on Build GRCh38
rs532134952 Chr.4: 119304361-119304361 on Build GRCh38
rs539557311 Chr.4: 119304353-119304353 on Build GRCh38
rs766331080 Chr.4: 119304284-119304284 on Build GRCh38

View Full Product Details