Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113425542-113425803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
UniGene
Hs.632691
Gene Chromosome Location
Chr.9: 113410644-113429684 on Build GRCh38
Transcripts
NM_001278629.1 NM_001278630.1 NM_152786.2 XM_005251885.3 XM_011518477.1 XM_011518478.1 XM_011518479.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4456217 c.1000C>T p.P334S Chr.9: 113425700-113425700 on Build GRCh38
5467571 c.994A>G p.K332E Chr.9: 113425694-113425694 on Build GRCh38
6616965 c.954G>T p.K318N Chr.9: 113425654-113425654 on Build GRCh38
3903234 c.1028A>G p.Y343C Chr.9: 113425728-113425728 on Build GRCh38
1496990 c.1012G>A p.V338I Chr.9: 113425712-113425712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772986146 Chr.9: 113425681-113425681 on Build GRCh38
rs996955379 Chr.9: 113425583-113425583 on Build GRCh38
rs898888624 Chr.9: 113425777-113425777 on Build GRCh38
rs962962889 Chr.9: 113425677-113425677 on Build GRCh38
rs573828696 Chr.9: 113425722-113425722 on Build GRCh38
rs917030361 Chr.9: 113425750-113425750 on Build GRCh38
rs142848142 Chr.9: 113425693-113425693 on Build GRCh38
rs763882417 Chr.9: 113425718-113425718 on Build GRCh38
rs746481551 Chr.9: 113425654-113425654 on Build GRCh38
rs780477872 Chr.9: 113425607-113425607 on Build GRCh38
rs781250277 Chr.9: 113425643-113425643 on Build GRCh38
rs770399149 Chr.9: 113425727-113425742 on Build GRCh38
rs759524254 Chr.9: 113425702-113425702 on Build GRCh38
rs768575638 Chr.9: 113425629-113425629 on Build GRCh38
rs145109883 Chr.9: 113425712-113425712 on Build GRCh38
rs371753726 Chr.9: 113425771-113425771 on Build GRCh38
rs778016915 Chr.9: 113425764-113425764 on Build GRCh38
rs761406961 Chr.9: 113425741-113425741 on Build GRCh38
rs926015206 Chr.9: 113425575-113425575 on Build GRCh38
rs747577139 Chr.9: 113425623-113425623 on Build GRCh38
rs138361218 Chr.9: 113425680-113425680 on Build GRCh38
rs267602092 Chr.9: 113425707-113425707 on Build GRCh38
rs751941724 Chr.9: 113425724-113425724 on Build GRCh38
rs771894456 Chr.9: 113425721-113425721 on Build GRCh38
rs748293105 Chr.9: 113425757-113425757 on Build GRCh38
rs745638713 Chr.9: 113425691-113425691 on Build GRCh38
rs375640602 Chr.9: 113425615-113425615 on Build GRCh38
rs1046346449 Chr.9: 113425703-113425703 on Build GRCh38
rs758911831 Chr.9: 113425604-113425604 on Build GRCh38
rs145630336 Chr.9: 113425662-113425662 on Build GRCh38
rs767080167 Chr.9: 113425711-113425711 on Build GRCh38
rs781231767 Chr.9: 113425751-113425751 on Build GRCh38
rs768060619 Chr.9: 113425598-113425598 on Build GRCh38
rs774211483 Chr.9: 113425773-113425773 on Build GRCh38
rs557768922 Chr.9: 113425585-113425585 on Build GRCh38
rs750782512 Chr.9: 113425720-113425720 on Build GRCh38
rs749545299 Chr.9: 113425768-113425768 on Build GRCh38
rs199691637 Chr.9: 113425746-113425746 on Build GRCh38
rs545206014 Chr.9: 113425692-113425692 on Build GRCh38
rs371997676 Chr.9: 113425763-113425763 on Build GRCh38

View Full Product Details