Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70452577-70452834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG3
Entrez Gene ID
1741
Gene Name
discs large MAGUK scaffold protein 3
Gene Aliases
MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR
UniGene
Hs.721586
Gene Chromosome Location
Chr.X: 70444850-70505490 on Build GRCh38
Transcripts
NM_020730.2 NM_021120.3 XM_005262248.3 XM_006724625.2 XM_006724626.2 XM_011530883.1 XM_017029322.1 XM_017029323.1 XM_017029324.1

SNP Detail

SNP ID Chromosome Location
rs769261082 Chr.X: 70452736-70452736 on Build GRCh38
rs781426250 Chr.X: 70452700-70452700 on Build GRCh38
rs746231348 Chr.X: 70452705-70452705 on Build GRCh38
rs771487436 Chr.X: 70452652-70452652 on Build GRCh38
rs954195677 Chr.X: 70452640-70452640 on Build GRCh38
rs759184230 Chr.X: 70452756-70452756 on Build GRCh38
rs766017171 Chr.X: 70452636-70452636 on Build GRCh38
rs771161683 Chr.X: 70452604-70452604 on Build GRCh38
rs758319980 Chr.X: 70452661-70452661 on Build GRCh38
rs749865823 Chr.X: 70452730-70452730 on Build GRCh38
rs753601803 Chr.X: 70452639-70452639 on Build GRCh38
rs767200604 Chr.X: 70452621-70452621 on Build GRCh38
rs762920372 Chr.X: 70452793-70452793 on Build GRCh38
rs780858258 Chr.X: 70452610-70452611 on Build GRCh38
rs765164328 Chr.X: 70452655-70452655 on Build GRCh38
rs765074594 Chr.X: 70452767-70452767 on Build GRCh38
rs1051718631 Chr.X: 70452754-70452754 on Build GRCh38
rs369619126 Chr.X: 70452687-70452687 on Build GRCh38
rs12010766 Chr.X: 70452805-70452805 on Build GRCh38
rs761408062 Chr.X: 70452614-70452614 on Build GRCh38
rs776898687 Chr.X: 70452606-70452606 on Build GRCh38
rs925984381 Chr.X: 70452672-70452672 on Build GRCh38
rs751612224 Chr.X: 70452802-70452802 on Build GRCh38
rs376966381 Chr.X: 70452674-70452674 on Build GRCh38
rs772761894 Chr.X: 70452634-70452634 on Build GRCh38
rs749457448 Chr.X: 70452622-70452622 on Build GRCh38
rs772630212 Chr.X: 70452737-70452737 on Build GRCh38
rs373735549 Chr.X: 70452692-70452692 on Build GRCh38
rs757449904 Chr.X: 70452693-70452693 on Build GRCh38
rs770445136 Chr.X: 70452607-70452607 on Build GRCh38
rs199928682 Chr.X: 70452669-70452669 on Build GRCh38
rs764051391 Chr.X: 70452801-70452801 on Build GRCh38
rs757057869 Chr.X: 70452606-70452607 on Build GRCh38
rs745750102 Chr.X: 70452652-70452652 on Build GRCh38
rs186178441 Chr.X: 70452741-70452741 on Build GRCh38
rs770212156 Chr.X: 70452723-70452723 on Build GRCh38
rs756114403 Chr.X: 70452605-70452606 on Build GRCh38
rs776283839 Chr.X: 70452742-70452742 on Build GRCh38
rs747255250 Chr.X: 70452602-70452602 on Build GRCh38
rs780557337 Chr.X: 70452725-70452725 on Build GRCh38
rs989042778 Chr.X: 70452770-70452770 on Build GRCh38
rs12850410 Chr.X: 70452612-70452612 on Build GRCh38
rs752699002 Chr.X: 70452660-70452660 on Build GRCh38
rs752468086 Chr.X: 70452777-70452777 on Build GRCh38

View Full Product Details