Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73730345-73730606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 XM_017010283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670334 c.507G>T p.K169N Chr.6: 73730574-73730574 on Build GRCh38
5453518 c.325C>T p.R109C Chr.6: 73730392-73730392 on Build GRCh38
1312384 c.298G>A p.E100K Chr.6: 73730365-73730365 on Build GRCh38
1697543 c.368C>T p.S123L Chr.6: 73730435-73730435 on Build GRCh38
5455519 c.488C>T p.S163F Chr.6: 73730555-73730555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370833000 Chr.6: 73730529-73730529 on Build GRCh38
rs754963873 Chr.6: 73730576-73730576 on Build GRCh38
rs183425587 Chr.6: 73730383-73730383 on Build GRCh38
rs145209100 Chr.6: 73730497-73730497 on Build GRCh38
rs187899427 Chr.6: 73730517-73730517 on Build GRCh38
rs552504094 Chr.6: 73730428-73730428 on Build GRCh38
rs764481178 Chr.6: 73730547-73730547 on Build GRCh38
rs147342140 Chr.6: 73730401-73730401 on Build GRCh38
rs111900767 Chr.6: 73730429-73730429 on Build GRCh38
rs544522678 Chr.6: 73730380-73730380 on Build GRCh38
rs762131867 Chr.6: 73730414-73730414 on Build GRCh38
rs750490627 Chr.6: 73730523-73730523 on Build GRCh38
rs752824497 Chr.6: 73730477-73730477 on Build GRCh38
rs868531522 Chr.6: 73730524-73730524 on Build GRCh38
rs972091957 Chr.6: 73730397-73730397 on Build GRCh38
rs534213077 Chr.6: 73730392-73730392 on Build GRCh38
rs768337926 Chr.6: 73730393-73730393 on Build GRCh38
rs751646436 Chr.6: 73730446-73730446 on Build GRCh38
rs746501980 Chr.6: 73730499-73730499 on Build GRCh38
rs1056234377 Chr.6: 73730431-73730431 on Build GRCh38
rs753645455 Chr.6: 73730377-73730377 on Build GRCh38
rs138148675 Chr.6: 73730521-73730521 on Build GRCh38
rs752703405 Chr.6: 73730496-73730497 on Build GRCh38
rs766306053 Chr.6: 73730562-73730562 on Build GRCh38
rs370528139 Chr.6: 73730396-73730396 on Build GRCh38
rs771448010 Chr.6: 73730518-73730518 on Build GRCh38
rs376274092 Chr.6: 73730426-73730426 on Build GRCh38
rs769447652 Chr.6: 73730515-73730515 on Build GRCh38
rs778795227 Chr.6: 73730493-73730493 on Build GRCh38
rs61730425 Chr.6: 73730549-73730549 on Build GRCh38
rs143082026 Chr.6: 73730381-73730381 on Build GRCh38
rs894715132 Chr.6: 73730453-73730456 on Build GRCh38
rs767750886 Chr.6: 73730460-73730460 on Build GRCh38
rs747748880 Chr.6: 73730513-73730513 on Build GRCh38
rs750252206 Chr.6: 73730496-73730496 on Build GRCh38
rs750225907 Chr.6: 73730548-73730548 on Build GRCh38
rs780508514 Chr.6: 73730582-73730582 on Build GRCh38
rs751409246 Chr.6: 73730573-73730573 on Build GRCh38
rs201515057 Chr.6: 73730389-73730389 on Build GRCh38
rs200235560 Chr.6: 73730378-73730378 on Build GRCh38
rs775728564 Chr.6: 73730532-73730532 on Build GRCh38
rs760918169 Chr.6: 73730539-73730539 on Build GRCh38
rs137899447 Chr.6: 73730512-73730512 on Build GRCh38
rs370649786 Chr.6: 73730485-73730485 on Build GRCh38
rs151159309 Chr.6: 73730388-73730388 on Build GRCh38
rs759584241 Chr.6: 73730450-73730450 on Build GRCh38
rs909072132 Chr.6: 73730390-73730390 on Build GRCh38
rs770102329 Chr.6: 73730424-73730424 on Build GRCh38

View Full Product Details