Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 66379277-66379520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA3
Entrez Gene ID
29119
Gene Name
catenin alpha 3
Gene Aliases
ARVD13, VR22
UniGene
Hs.21375
Gene Chromosome Location
Chr.10: 65912518-67696217 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 XM_017016157.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4649457 c.1538A>G p.H513R Chr.10: 66379346-66379346 on Build GRCh38
4412617 c.1542C>A p.I514I Chr.10: 66379342-66379342 on Build GRCh38
2158776 c.1570G>T p.A524S Chr.10: 66379314-66379314 on Build GRCh38
4468291 c.1536C>T p.S512S Chr.10: 66379348-66379348 on Build GRCh38
3686820 c.1603C>T p.R535C Chr.10: 66379281-66379281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777498783 Chr.10: 66379378-66379378 on Build GRCh38
rs375914255 Chr.10: 66379480-66379480 on Build GRCh38
rs369650158 Chr.10: 66379310-66379310 on Build GRCh38
rs765560825 Chr.10: 66379325-66379325 on Build GRCh38
rs532775825 Chr.10: 66379391-66379391 on Build GRCh38
rs1035750089 Chr.10: 66379453-66379453 on Build GRCh38
rs1002736517 Chr.10: 66379452-66379452 on Build GRCh38
rs760926013 Chr.10: 66379302-66379302 on Build GRCh38
rs377413642 Chr.10: 66379345-66379345 on Build GRCh38
rs766151100 Chr.10: 66379486-66379486 on Build GRCh38
rs868277987 Chr.10: 66379317-66379317 on Build GRCh38
rs903000291 Chr.10: 66379443-66379443 on Build GRCh38
rs892579657 Chr.10: 66379462-66379462 on Build GRCh38
rs771313986 Chr.10: 66379378-66379379 on Build GRCh38
rs1050514681 Chr.10: 66379351-66379351 on Build GRCh38
rs746368312 Chr.10: 66379397-66379398 on Build GRCh38
rs12761828 Chr.10: 66379384-66379384 on Build GRCh38
rs780966240 Chr.10: 66379341-66379341 on Build GRCh38
rs771660886 Chr.10: 66379386-66379386 on Build GRCh38
rs777145148 Chr.10: 66379381-66379382 on Build GRCh38
rs756034227 Chr.10: 66379353-66379353 on Build GRCh38
rs770508772 Chr.10: 66379383-66379383 on Build GRCh38
rs546606277 Chr.10: 66379421-66379421 on Build GRCh38
rs373151978 Chr.10: 66379335-66379335 on Build GRCh38
rs747411476 Chr.10: 66379316-66379318 on Build GRCh38
rs920257424 Chr.10: 66379368-66379368 on Build GRCh38
rs775830635 Chr.10: 66379387-66379387 on Build GRCh38
rs754275515 Chr.10: 66379314-66379314 on Build GRCh38
rs762067610 Chr.10: 66379316-66379316 on Build GRCh38
rs372877107 Chr.10: 66379405-66379405 on Build GRCh38
rs371149404 Chr.10: 66379457-66379457 on Build GRCh38
rs1012881526 Chr.10: 66379487-66379487 on Build GRCh38
rs893024601 Chr.10: 66379376-66379376 on Build GRCh38
rs758592447 Chr.10: 66379448-66379456 on Build GRCh38
rs1041694409 Chr.10: 66379423-66379423 on Build GRCh38
rs144269113 Chr.10: 66379326-66379326 on Build GRCh38
rs373940157 Chr.10: 66379382-66379382 on Build GRCh38

View Full Product Details